Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

153591

Sigma-Aldrich

Diphenylcarbamoyl chloride

98%

Synonym(s):

Chloroformic acid diphenylamide, Diphenylcarbamic chloride, NSC 6788

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)2NCOCl
CAS Number:
Molecular Weight:
231.68
Beilstein:
515312
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

powder

solubility

organic solvents: soluble

functional group

amine

SMILES string

ClC(=O)N(c1ccccc1)c2ccccc2

InChI

1S/C13H10ClNO/c14-13(16)15(11-7-3-1-4-8-11)12-9-5-2-6-10-12/h1-10H

InChI key

XNBKKRFABABBPM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Diphenylcarbamoyl chloride was used to isolate peptido-keratan sulphate fragments from bovine intervertebral discs.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B - Skin Sens. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

J M Dickenson et al.
The Biochemical journal, 282 ( Pt 1), 267-271 (1992-02-15)
Peptido-keratan sulphate fragments were isolated from the nucleus pulposus of bovine intervertebral discs (2-year-old animals) after digestion with chondroitin ABC lyase followed by digestion with diphenylcarbamoyl chloride-treated trypsin of A1D1 proteoglycans and gel-permeation chromatography on Sepharose CL-6B. The peptido-keratan sulphate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service