Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

259950

Sigma-Aldrich

Benzoyl chloride

99%

Synonym(s):

Benzoic acid chloride, alpha-chlorobenzaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5COCl
CAS Number:
Molecular Weight:
140.57
Beilstein:
471389
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21
Pricing and availability is not currently available.

vapor density

4.88 (vs air)

Quality Level

vapor pressure

1 mmHg ( 32 °C)

Assay

98.0-100.5%
99%

form

liquid

autoignition temp.

1056 °F

expl. lim.

4.9 %

impurities

≤0.002% P compounds

ign. residue

≤0.005%

refractive index

n20/D 1.553 (lit.)

bp

198 °C (lit.)

mp

−1 °C (lit.)

transition temp

freezing point −2.0-0.0 °C

density

1.211 g/mL at 25 °C (lit.)

cation traces

Fe: ≤0.001%
heavy metals (as Pb): ≤0.001%

functional group

acyl chloride
phenyl

SMILES string

ClC(=O)c1ccccc1

InChI

1S/C7H5ClO/c8-7(9)6-4-2-1-3-5-6/h1-5H

InChI key

PASDCCFISLVPSO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Benzoyl chloride may be used in the synthesis of the following organic building blocks:
  • N,N-diethylbenzamide via condensation with diethylamine[1]
  • N-2-bromophenylbenzamide by reacting with 2-bromoaniline via N-benzoylation[2]
  • propargyl benzoate via O-benzoylation of propargyl alcohol[3]

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

161.6 °F - closed cup

Flash Point(C)

72 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation of (E)-N,N-Diethyl-2-styrylbenzamide by Rh-Catalyzed C-H Activation
Patureau FW, et al
Organic Syntheses, 90, 41-41 (2013)
Ligand-free copper(II) oxide nanoparticles catalyzed synthesis of substituted benzoxazoles
Prasenjit S, et al
Organic Syntheses, 88, 398-398 (2011)
2-substituted-1,3-cyclohexadienes by intermolecular, methylene-free tandem enyne metathesis
Kulkarni AA and Diver ST
Organic Syntheses, 83, 200-200 (2006)
Solid catalyst treated with anion: XVIII. Benzoylation of toluene with benzoyl chloride and benzoic anhydride catalysed by solid superacid of sulfate-supported alumina.
Arata K and Hino M.
Applied Catalysis, 59(1), 197-204 (1990)
High activity of iron containing metal-organic-framework in acylation of p-xylene with benzoyl chloride.
Kurfirtova L, et al.
Catalysis Today, 179(1), 85-90 (2012)

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service