Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D152803

Sigma-Aldrich

Dimethylcarbamyl chloride

98%

Synonym(s):

Chloroformic acid dimethyl amide, Dimethylcarbamoyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2NCOCl
CAS Number:
Molecular Weight:
107.54
Beilstein:
878197
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.453 (lit.)

bp

167-168 °C/775 mmHg (lit.)

mp

−33 °C (lit.)

density

1.168 g/mL at 25 °C (lit.)

SMILES string

CN(C)C(Cl)=O

InChI

1S/C3H6ClNO/c1-5(2)3(4)6/h1-2H3

InChI key

YIIMEMSDCNDGTB-UHFFFAOYSA-N

Application

Dimethylcarbamyl chloride can be used to synthesize:
  • Disubstituted carbamates from benzylphenols.
  • 2-Cyanoisonicotinamide by reacting with isonicotinic acid N-oxide and zinc cyanide. This method was adopted to synthesize a novel xanthine oxidoreductase inhibitor.
  • Acetylcholinesterase (AChE) and serotonin transporter (SERT) dual inhibitors.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 1B - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

179.6 °F - closed cup

Flash Point(C)

82 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Design and synthesis of dual inhibitors of acetylcholinesterase and serotonin transporter targeting potential agents for Alzheimer's disease.
Kogen H, et al.
Organic Letters, 4(20), 3359-3362 (2002)
Dimethylcarbamoyl chloride.
Report on carcinogens : carcinogen profiles, 10, 106-107 (2004-08-25)
S J Garte et al.
Environmental health perspectives, 81, 29-31 (1989-05-01)
Techniques of molecular biology have been used to determine the relationship of cellular oncogenes to mechanisms of experimental carcinogenesis. Model systems involving three direct-acting alkylating carcinogens, two organ sites, and two species have been employed to elucidate the relationships between
Dimethylcarbamoyl chloride.
IARC monographs on the evaluation of carcinogenic risks to humans, 71 Pt 2, 531-543 (1999-09-07)
C A Snyder et al.
Archives of toxicology, 61(1), 3-6 (1987-01-01)
A new inhalation exposure system has been developed which allows the determination of inhaled doses of vapors and gases by laboratory rats. The system consists of saran bags connected to head-only exposure cylinders via one-way valves. One bag serves as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service