Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

103918

Sigma-Aldrich

2-Chlorobenzoyl chloride

95%

Synonym(s):

o-Chlorobenzoyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4COCl
CAS Number:
Molecular Weight:
175.01
Beilstein:
386435
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

refractive index

n20/D 1.572 (lit.)

bp

238 °C (lit.)

mp

−4-−3 °C (lit.)

density

1.382 g/mL at 25 °C (lit.)

SMILES string

ClC(=O)c1ccccc1Cl

InChI

1S/C7H4Cl2O/c8-6-4-2-1-3-5(6)7(9)10/h1-4H

InChI key

ONIKNECPXCLUHT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Chlorobenzoyl chloride reacts with aromatic amines and ammonium thiocyanate using polyethylene glycol-400 as the catalyst under the condition of solid-liquid phase-transfer catalysis to form N-aryl-N′(2-chlorobenzoyl) thioureas. 2-Chlorobenzoyl chloride causes the acylation of polystyrene during the preparation and regeneration of the polystyrene-based resin.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

255.2 °F - closed cup - DIN 51758

Flash Point(C)

124 °C - closed cup - DIN 51758

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Simple and efficient synthesis of 2-chlorotritylchloride resin.
Orosz G and Kiss L.
Tetrahedron Letters, 39(20), 3241-3242 (1998)
Phase-transfer-catalyzed synthesis of N-aryl-N'-(2-chlorobenzoyl)-thiourea derivatives.
Zhang Y, et al.
Synthetic Communications, 27(5), 751-756 (1997)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service