Skip to Content

1197007

USP

Diethyltoluamide

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

N,N-Diethyl-3-methylbenzamide, N,N-Diethyl-m-toluamide, m-Toluic acid diethylamide, DEET

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
CH3C6H4CON(C2H5)2
CAS Number:
Molecular Weight:
191.27
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
2046711
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

vapor density

6.7 (vs air)

vapor pressure

<0.01 mmHg ( 25 °C)

API family

diethyltoluamide

manufacturer/tradename

USP

refractive index

n20/D 1.523 (lit.)

bp

111 °C/1 mmHg, 160 °C/19 mmHg (lit.)

density

0.998 g/mL at 20 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CCN(CC)C(=O)c1cccc(C)c1

InChI

1S/C12H17NO/c1-4-13(5-2)12(14)11-8-6-7-10(3)9-11/h6-9H,4-5H2,1-3H3

InChI key

MMOXZBCLCQITDF-UHFFFAOYSA-N

General description

Find answers to frequently asked questions on USP products in USP FAQs.
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Diethyltoluamide USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Diethyltoluamide Topical Solution

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.


Still not finding the right product?

Explore all of our products under Diethyltoluamide


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

203.0 °F - closed cup

flash_point_c

95 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library