Merck
V800166
Sigma-Aldrich
Diisopropyl ether
LR, 98%
Sinónimos:
Isopropyl ether
About This Item
Fórmula lineal:
(CH3)2CHOCH(CH3)2
Número de CAS:
Peso molecular:
102.17
Beilstein/REAXYS Number:
1731256
EC Number:
MDL number:
UNSPSC Code:
12191502
PubChem Substance ID:
grade
LR
vapor density
3.5 (vs air)
vapor pressure
120 mmHg ( 20 °C)
product line
Vetec
assay
98%
form
liquid
autoignition temp.
827 °F
expl. lim.
1-21 %, 100 °F
refractive index
n20/D 1.367 (lit.)
bp
68-69 °C (lit.)
mp
−85 °C (lit.)
density
0.725 g/mL at 25 °C (lit.)
SMILES string
CC(C)OC(C)C
InChI
1S/C6H14O/c1-5(2)7-6(3)4/h5-6H,1-4H3
InChI key
ZAFNJMIOTHYJRJ-UHFFFAOYSA-N
Legal Information
Vetec is a trademark of Merck KGaA, Darmstadt, Germany
pictograms
FlameExclamation mark
signalword
Danger
hcodes
Hazard Classifications
Flam. Liq. 2 - STOT SE 3
target_organs
Central nervous system
supp_hazards
Storage Class
3 - Flammable liquids
wgk_germany
WGK 1
flash_point_f
-20.2 °F - closed cup
flash_point_c
-29 °C - closed cup
Visite la Librería de documentos
P Adlercreutz
Biochimica et biophysica acta, 1163(2), 144-148 (1993-05-13)
Horse liver alcohol dehydrogenase and alpha-chymotrypsin were deposited on a porous support material, Celite. After equilibration at a well-defined water activity, the catalytic activity was measured with diisopropyl ether as reaction medium. The effects of the presence of polyols and
R Gonzalez-Olmos et al.
Chemosphere, 71(11), 2098-2105 (2008-02-27)
Methyl tert-butyl ether (MTBE) is the most widely used oxygenate in gasoline blending and has become one of the world's most widespread groundwater and surface water pollutants. Alternative oxygenates to MTBE, namely ethyl tert-butyl ether (ETBE), tert-amyl ether (TAME) and
B E Cham et al.
Journal of virological methods, 137(1), 160-163 (2006-07-01)
Many viruses including HIV, hepatitis C and hepatitis B, have an outer lipid envelope which maintains inserted viral peptides in the "correct" functional conformation and orientation. Disruption of the lipid envelope by most solvents destroys infectivity and often results in
R van Gijn et al.
Journal of chromatography. B, Biomedical applications, 667(2), 269-276 (1995-05-19)
The staurosporine derivative, N-benzoylstaurosporine (CGP 41 251; I), is a protein kinase C inhibitor that has been selected for phase I clinical evaluation in cancer patients. We have developed a selective and sensitive assay of the drug and three potential
A Hickel et al.
Biotechnology and bioengineering, 65(4), 425-436 (1999-10-03)
A novel recycle reactor has been designed to determine the interfacial activity of hydroxynitrile lyase in a diisopropyl ether (DIPE)/water two-phase system. The reactor provides a known interfacial area. Enzyme activity toward mandelonitrile cleavage is continuously measured in the reactor