Documentos clave
Z742778
smartPans cahn style aluminim weighing dish
pack of 20 (Dishes supplied in flip top vial)
Seleccione un Tamaño
20 EA
US$ 210,00
US$ 210,00
PRECIOS SIN IMPUESTOS NACIONALES
Check Cart for Availability
Seleccione un Tamaño
Cambiar Vistas
20 EA
US$ 210,00
About This Item
Código UNSPSC:
41121812
NACRES:
NB.43
US$ 210,00
PRECIOS SIN IMPUESTOS NACIONALES
Check Cart for Availability
Materiales
aluminum
envase
pack of 20 (Dishes supplied in flip top vial)
fabricante / nombre comercial
LevGo Inc 41568-020
Diám. × H × grosor
25 mm × 3.59 mm × 0.038 mm
peso
53 mgEach boat
Descripción general
Ideal for microbalances, organic micro elementary / thermal analysis, and pharmaceutical use.
  • Made of Ultra High Vacuum (UVH) Foil - “A” wettable and completely free from oils normally found on other grades of aluminum foil.
  • Foil meets ASTM B-479, specification 3.1.4 and 10.3.1.
  • “Cahn style” accessory pan / disc format
  • Compatible with most commercially available analyzers and microbalances
  • Foil and Pans are made in the USA
  • Packaged 20 or 100 per easy-to-use PP flip top vials - Both the pans and the vials recyclable
Información legal
smartPans is a trademark of LevGo Inc.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)