Documentos clave
Z290475
Sigma-Aldrich
Sigma-Aldrich® vacuum filtration assembly
for 25 mm filters with glass support, #5 stopper, same as Z290467 but without filtration flask
Seleccione un Tamaño
1 EA
US$ 414,00
US$ 414,00
PRECIOS SIN IMPUESTOS NACIONALES
Check Cart for Availability
Seleccione un Tamaño
Cambiar Vistas
1 EA
US$ 414,00
About This Item
Código UNSPSC:
41104900
NACRES:
NB.24
US$ 414,00
PRECIOS SIN IMPUESTOS NACIONALES
Check Cart for Availability
Materiales
glass housing
glass membrane
Características
cap plug seal
graduation
envase
pkg of 1 ea
embudo capacidad
15 mL
cuello diám.
27 mm
Descripción general
Same as Z290467 but without filtration flask.
Vacuum filtration apparatus for HPLC solvent purification, particulate analysis, bacteriological studies, radiochemical determinations, quantitative gravimetric analysis, and similar applications. Clamp seals filter membrane between flanges on funnel top and funnel stem.
Includes: Funnel top, funnel stem/membrane support, aluminum clamp, stopper (except standard taper units) and filtration flask (where indicated).
For replacement parts, visit our online glassware catalog at www.sigma-aldrich.com
Información legal
SIGMA-ALDRICH is a registered trademark of Merck KGaA, Darmstadt, Germany
pieza de repuesto
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)