245119
Reagent Alcohol
reagent, spectrophotometric grade, denatured, suitable for UV/Vis spectroscopy, contains 5% isopropyl alcohol and 5% methyl alcohol as denaturant
Sinónimos:
Ethanol, RGA, denatured alcohol
About This Item
Productos recomendados
product name
Reagent Alcohol, spectrophotometric grade
grade
denatured
reagent
spectrophotometric grade
vapor density
1.59 (vs air)
form
liquid
contains
5% isopropyl alcohol as denaturant
5% methyl alcohol as denaturant
expl. lim.
3.1-27.7 %
technique(s)
UV/Vis spectroscopy: suitable
impurities
≤0.10% water
evapn. residue
<0.0005%
bp
78 °C
mp
-114 °C
density
0.785 g/mL at 25 °C
λ
H2O reference
UV absorption
λ: 205 nm Amax: 1.00
λ: 210 nm Amax: 0.65
λ: 220 nm Amax: 0.35
λ: 255 nm Amax: 0.04
λ: 300-400 nm Amax: 0.01
InChI
1S/C2H6O/c1-2-3/h3H,2H2,1H3
InChI key
LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Components
greener alternative product
signalword
Danger
hcodes
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 2
target_organs
Eyes,Central nervous system
Storage Class
3 - Flammable liquids
wgk_germany
WGK 2
flash_point_f
48.2 °F - closed cup
flash_point_c
9 °C - closed cup