Documentos clave
Z678325
Finnpipette® F2 variable volume pipette
volume 10-100 μL
Seleccione un Tamaño
1 EA
US$ 617,00
US$ 617,00
PRECIOS SIN IMPUESTOS NACIONALES
Check Cart for Availability
Seleccione un Tamaño
Cambiar Vistas
1 EA
US$ 617,00
About This Item
Código UNSPSC:
41121500
NACRES:
NB.01
US$ 617,00
PRECIOS SIN IMPUESTOS NACIONALES
Check Cart for Availability
Características
color-coded yellow
electronic: no
single-channel
envase
pack of 1 ea
fabricante / nombre comercial
Finn 4642070
volumen
10-100 μL
Descripción general
Finnpipette F2 has a light pipetting action, is easy to use and is fully autoclavable. Accuracy and precision are the most vital aspect of any pipette so the Finnpipette F2 was developed with Advanced Volume Gearing (AVG). This is a self-supporting modular volume adjustment mechanism separated from the pipette body. This improves accuracy and precision by preventing possible effects of hand warmth on the accuracy of measurements.
  • Advanced Volume Gearing mechanism (AVG) ensures increased accuracy and precision
  • Big display with white numbers on black background for increased visibility
  • Double action pipetting button with very light and smooth pipetting action
  • Fully autoclavable
  • Super blow-out piston with volumes of 50μl and below
Información legal
Finnpipette is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)