Documentos clave
Z678139
Finnpipette® F3 variable volume pipette
volume 20-200 μL
Seleccione un Tamaño
1 EA
US$ 569,00
US$ 569,00
PRECIOS SIN IMPUESTOS NACIONALES
Check Cart for Availability
Seleccione un Tamaño
Cambiar Vistas
1 EA
US$ 569,00
About This Item
Código UNSPSC:
41121500
NACRES:
NB.01
US$ 569,00
PRECIOS SIN IMPUESTOS NACIONALES
Check Cart for Availability
Características
color-coded yellow
electronic: no
single-channel
envase
pack of 1 ea
fabricante / nombre comercial
Finn 4640050
volumen
20-200 μL
Categorías relacionadas
Descripción general
Finnpipette F3 has a double action pipetting button with rotating upper part enabling smooth and steady pipetting action at the same time as preventing accidental volume alterations. Finpipette F3 is equipped with the Advanced Volume Gearing (AVG). This is a self-supporting modular volume adjustment mechanism separated from the pipette body. This improves accuracy and precision by preventing possible effects of hand warmth on the accuracy of measurements.
  • Double action pipetting button with very light and smooth pipetting action
  • Advanced Volume Gearing mechanism (AVG) ensures increased accuracy and precision
  • Big display with white numbers on black background for increased visibility
  • Color coded for easy tip volume recognition
Información legal
Finnpipette is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)