Z276596
Gas cylinder hand truck
no. of cylinders: 1
About This Item
Código UNSPSC:
41100000
NACRES:
NB.51
Productos recomendados
Altura
1067 mm
42 in.
peso
45 lb
Descripción general
These collapsible trucks make cylinder handling safe and convenient. Adjustable, spring-locked carriages permit cylinders to be carried at a comfortable angle (45°). Trucks have painted tubular steel frames, cylinder restraining straps, and pneumatic wheels/casters.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)