Skip to Content
Merck
All Photos(1)

Key Documents

442716

Supelco

Naphthalene-d8

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10D8
CAS Number:
Molecular Weight:
136.22
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

analytical standard

vapor density

4.4 (vs air)

vapor pressure

0.03 mmHg ( 25 °C)

CofA

current certificate can be downloaded

expl. lim.

5.9 %

packaging

ampule of 250 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

218 °C (lit.)

mp

80-82 °C (lit.)

application(s)

environmental

format

neat

mass shift

M+8

storage temp.

2-30°C

SMILES string

[2H]c1c([2H])c([2H])c2c([2H])c([2H])c([2H])c([2H])c2c1[2H]

InChI

1S/C10H8/c1-2-6-10-8-4-3-7-9(10)5-1/h1-8H/i1D,2D,3D,4D,5D,6D,7D,8D

InChI key

UFWIBTONFRDIAS-PGRXLJNUSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Flam. Sol. 2

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

174.2 °F - closed cup

Flash Point(C)

79.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amanda Kowalsick et al.
Journal of chromatography. A, 1370, 230-239 (2014-12-03)
Seasonal variation in tea (Camellia sinensis (L.) Kuntze; Theaceae) chemistry was investigated using automated sequential, multidimensional gas chromatography/mass spectrometry (GC-GC/MS). Metabolite libraries were produced for teas harvested from the Bulang Mountains in Yunnan, China before and after the onset of
Zoe Ruge et al.
Environmental science & technology, 49(23), 13777-13786 (2015-10-06)
Polycyclic aromatic hydrocarbons (PAHs) and polybrominated diphenylethers (PBDEs) are both currently released into the environment from anthropogenic activity. Both are hence primarily associated with populated or industrial areas, although wildfires can be an important source of PAHs, as well. Polyethylene
Badreddine Barhoumi et al.
Marine environmental research, 101, 184-195 (2014-08-12)
In this study, biological responses and contaminant levels in biological tissues were investigated in grass goby fish specimens (Zosterisessor ophiocephalus) collected from five stations in a moderately polluted ecosystem, namely the Bizerte lagoon on the north coast of Tunisia. The
Askin Birgul et al.
Archives of environmental contamination and toxicology, 68(1), 46-63 (2014-12-07)
Ambient air polycyclic aromatic hydrocarbon (PAH) samples were collected at traffic, residential, coastal, and semiurban sites in Bursa, Turkey, between June 2008 and June 2009. For the traffic, residential, coastal, and semiurban sites, the average gas phase total PAH (∑12PAH)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service