Merck
  • Pollution biomonitoring in the Bizerte lagoon (Tunisia), using combined chemical and biomarker analyses in grass goby, Zosterisessor ophiocephalus (Teleostei, Gobiidae).
Pollution biomonitoring in the Bizerte lagoon (Tunisia), using combined chemical and biomarker analyses in grass goby, Zosterisessor ophiocephalus (Teleostei, Gobiidae).
Marine environmental research (2014-08-12)
Badreddine Barhoumi, Christelle Clérandeau, Pierre-Yves Gourves, Karyn Le Menach, Yassine El Megdiche, Laurent Peluhet, Hélène Budzinski, Magalie Baudrimont, Mohamed Ridha Driss, Jérôme Cachot
ABSTRACT
In this study, biological responses and contaminant levels in biological tissues were investigated in grass goby fish specimens (Zosterisessor ophiocephalus) collected from five stations in a moderately polluted ecosystem, namely the Bizerte lagoon on the north coast of Tunisia. The following biomarkers were measured: muscular acetylcholinesterase (AChE), hepatic ethoxyresorufin-O-deethylase (EROD), glutathione-S-transferase (GST), catalase (CAT), lipoperoxidation (TBARS), condition factor (CF), and hepatosomatic index (HSI). These measurements were taken in parallel with the content of Organochlorine pesticides (OCPs), Polychlorinated biphenyls (PCBs), Polycyclic aromatic hydrocarbons (PAHs) and trace metals (As, Cr, Cu, Mn, Pb, V, Zn, Ag, Cd, Co and Ni) in muscle tissue. Total PAH concentrations ranged from 20.09 ± 0.68 to 105.77 ± 42.58 ng g(-1) dw, PCB from 33.19 ± 6.25 to 126.28 ± 7.37 ng g(-1) dw, OCP from 11.26 ± 1.62 to 19.17 ± 2.06 ng g(-1) dw, and metals from 107.83 ± 1.83 to 187.21 ± 2.00 mg/kg dw. The highest levels of pollutants and biomarkers were observed at station S1, located in the Bizerte channel. Elevated EROD, GST and CAT activities, as well as TBARS levels in liver were positively correlated with tissue contaminant levels at station S1. Significant negative correlations were also found between hexachlorobenzene (HCB) and dichlorodiphenyltrichloroethane (DDTs) body burden with AChE activity in muscle at station S2. The integration of biological responses and contaminant tissue content indicated that certain areas of the Bizerte lagoon, notably station S1, are significantly impacted by various human activities, which likely represent a threat for aquatic wildlife. On the basis of these results, and due to its ecological characteristics, the grass goby appears a suitable indicator species for pollution biomonitoring in coastal marine areas along the Mediterranean Sea.
Sigma-Aldrich
Dibenzothiophene-d8, 98 atom % D, 98% (CP)
Sigma-Aldrich
Benzo[e]pyrene-d12, 98 atom % D, 98% (CP)
Supelco
Acenaphthylene, analytical standard
Sigma-Aldrich
Nitric-14N acid solution, ~10 N in H2O, 99.99 atom % 14N
Supelco
Aluminum oxide, activated, neutral, Brockmann Activity I
Supelco
Dibenzothiophene, analytical standard
Supelco
Acetone, analytical standard
Sigma-Aldrich
Naphthalene-d8, 99 atom % D, ≥98% (CP)
Sigma-Aldrich
Chrysene-d12, 98 atom % D
Sigma-Aldrich
Aluminum oxide, mesoporous, MSU-X (wormhole), average pore size 3.8 nm
Sigma-Aldrich
Dibenzothiophene, 98%
Sigma-Aldrich
Acenaphthylene, 75%
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Dibenzothiophene, ≥99%
Sigma-Aldrich
Benzo[a]pyrene-d12, 98 atom % D
Sigma-Aldrich
Acenaphthylene, 99%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Supelco
4-tert-Octylphenol monoethoxylate solution, 10 μg/mL in acetone, analytical standard
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99% (GC)
Sigma-Aldrich
Acetone, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
USP
Acetone, United States Pharmacopeia (USP) Reference Standard
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.8%