Skip to Content
Merck
All Photos(1)

Key Documents

456292

Sigma-Aldrich

Fluoranthene-d10

98 atom % D

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$203.00
100 MG
$263.00

$203.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

Select a Size

Change View
50 MG
$203.00
100 MG
$263.00

About This Item

Empirical Formula (Hill Notation):
C16D10
CAS Number:
Molecular Weight:
212.31
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.12

$203.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

isotopic purity

98 atom % D

Quality Level

Assay

99% (CP)

form

solid

bp

384 °C (lit.)

mp

110-113 °C (lit.)

application(s)

electronics

mass shift

M+10

SMILES string

[2H]c1c([2H])c([2H])c-2c(c1[2H])-c3c([2H])c([2H])c([2H])c4c([2H])c([2H])c([2H])c-2c34

InChI

1S/C16H10/c1-2-8-13-12(7-1)14-9-3-5-11-6-4-10-15(13)16(11)14/h1-10H/i1D,2D,3D,4D,5D,6D,7D,8D,9D,10D

InChI key

GVEPBJHOBDJJJI-LHNTUAQVSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

May be used as an analytical standard

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

388.4 °F - closed cup

Flash Point(C)

198.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Ahmad Omar-Ali et al.
Marine environmental research, 108, 33-44 (2015-05-10)
Alligator gar Atractosteus spatula acclimated to brackish water (9 ppt) were exposed to water accommodated fraction oil loadings (surrogate to Macondo Deepwater Horizon, northern Gulf of Mexico) of 0.5 and 4.0 gm oil/L tank water for 48 h. The surrogate

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service