Merck
  • Concentrations, gas-particle partitioning, and seasonal variations of polycyclic aromatic hydrocarbons at four sites in Turkey.
Concentrations, gas-particle partitioning, and seasonal variations of polycyclic aromatic hydrocarbons at four sites in Turkey.
Archives of environmental contamination and toxicology (2014-12-07)
Askin Birgul, Yücel Tasdemir
ABSTRACT
Ambient air polycyclic aromatic hydrocarbon (PAH) samples were collected at traffic, residential, coastal, and semiurban sites in Bursa, Turkey, between June 2008 and June 2009. For the traffic, residential, coastal, and semiurban sites, the average gas phase total PAH (∑12PAH) concentrations were 113 ± 131, 142 ± 204, 53 ± 73, and 19 ± 34 ng/m(3), respectively, whereas the average particle phase total PAH concentrations were 28 ± 36, 56 ± 85, 24 ± 40, and 11 ± 23 ng/m(3), respectively. Phenanthrene and fluoranthene had the highest concentrations of all of the sampling sites in the gas phase. The PAH concentrations in the heating period were 5-7 times greater than the nonheating period concentrations. Principal component analysis (PCA) was used to investigate the relationship between the levels of PAHs determined in ambient samples and their possible sources. The PCA model shows that coal combustion and vehicle emissions affected PAH emissions. Moreover, the molecular diagnostic ratios indicated that coal-burning and traffic emissions were the dominant PAH sources. The multiple linear regression analysis indicated that the meteorological parameters also affected the ambient PAH concentrations. The sampling site characteristics, meteorological conditions, dispersion, and local sources all affected the concentration levels.
Sigma-Aldrich
Methanol, NMR reference standard
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Acetone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Naphthalene-d8, 99 atom % D, ≥98% (CP)
Sigma-Aldrich
Acenaphthylene, 99%
Supelco
Methanol, analytical standard
Supelco
Acetone, analytical standard
Sigma-Aldrich
Acenaphthene-d10, 99 atom % D
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Acetone, natural, ≥97%
Supelco
Fluorene, analytical standard
Sigma-Aldrich
Acenaphthylene, 75%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Fluorene, 98%
Supelco
Acenaphthylene, analytical standard
Sigma-Aldrich
Acetone, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99% (GC)
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Acetone, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%