Skip to Content
Merck
All Photos(1)

Key Documents

Z657972

Hettich® centrifuge rotors

for ROTOFIX 32A and UNIVERSAL 320/320R Centrifuges, Swing-out Rotor, 6 place, to hold, 6 x 50 mL tubes with screw caps

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$1,530.00

$1,530.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
1 EA
$1,530.00

About This Item

UNSPSC Code:
41103900
NACRES:
NB.22

$1,530.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

feature

to hold 6 x 50 mL tubes with screw caps

packaging

pack of 1 ea

manufacturer/tradename

Hettich 1619

parameter

2,701 max. RCF

technique(s)

centrifugation: suitable

compatibility

for use with centrifuge models Universal 320/320R and Rotafix 32A

Looking for similar products? Visit Product Comparison Guide

Legal Information

Hettich is a registered trademark of Andreas Hettich GmbH & Co. KG

required but not provided

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service