Key Documents
Z601527
Hettich® centrifuge rotors
for UNIVERSAL 320/320R Centrifuge, Swing-out Rotor, 4 place
Select a Size
1 EA
$1,050.00
$1,050.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$1,050.00
About This Item
UNSPSC Code:
41103900
NACRES:
NB.78
$1,050.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
feature
to hold 4 x Suspensions (Models 1345, 1346, 1366, 1369, 1370, 1372, 1741, 1742, 1745, 1746)
packaging
pack of 1 ea
manufacturer/tradename
Hettich 1624
parameter
2,665 max. RCF
technique(s)
centrifugation: suitable
compatibility
for use with centrifuge models Universal 320/320R and Rotofix 32A
General description
The 4-place rotor 1624 (Z601527) is best suited for laboratories not exclusively involved with cytological examinations. Both tubes and cytological prepartions can be centrifuged without requiring a rotor change. Changing the buckets/carriers is all that is needed. Rotor 1624 (Z601527) accommodates cyto bucket/carriers 1660 and 1680.
Legal Information
Hettich is a registered trademark of Andreas Hettich GmbH & Co. KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)