Skip to Content
Merck
All Photos(1)

Key Documents

Z731773

Hettich® swing-out rotor, 4 place

for UNIVERSAL 320/320R Centrifuge, Swing-out Rotor, 4 place

Synonym(s):

Hettich® centrifuge rotors

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$1,180.00

$1,180.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
1 EA
$1,180.00

About This Item

UNSPSC Code:
41103900
NACRES:
NB.22

$1,180.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

feature

capacity variable (depends on adapter/bucket combination)
to hold 4 x 1452, 1660, 1680 buckets
Rotor only, does not include carriers (illustrated with carriers Hettich, 1427)

packaging

pack of 1 ea

manufacturer/tradename

Hettich 1494

parameter

4,193 max. RCF

technique(s)

centrifugation: suitable

compatibility

for use with centrifuge models Universal 320/320R

Looking for similar products? Visit Product Comparison Guide

Related Categories

Legal Information

Hettich is a registered trademark of Andreas Hettich GmbH & Co. KG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service