Key Documents
Z623598
Hettich® centrifuge adapters
for ROTINA 38/38R, ROTINA 420/420R and ROTANTA 460/460R, for 4.5-5 mL blood/urine tubes (11x92 mm) and 7 mL round bottom tubes (12x100 mm)
Select a Size
8 EA
$611.00
$611.00
Estimated to ship onApril 22, 2025
Select a Size
Change View
8 EA
$611.00
About This Item
UNSPSC Code:
41103911
NACRES:
NB.82
$611.00
Estimated to ship onApril 22, 2025
feature
Holds 7 mL tubes
packaging
pack of 8 ea
manufacturer/tradename
Hettich 5247
technique(s)
centrifugation: suitable
compatibility
for use with 5051 (buckets)
General description
  • compatible with bucket/carriers 5051 and 5628
Legal Information
Hettich is a registered trademark of Andreas Hettich GmbH & Co. KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)