Skip to Content
Merck
All Photos(1)

Key Documents

SML3824

Sigma-Aldrich

DRB18

≥98% (HPLC)

Synonym(s):

5,5′-(((4-Chloro-1,2-phenylene)bis(azanediyl))bis(methylene))bis(2-methylphenol), DRB 18, DRB-18

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H23ClN2O2
CAS Number:
Molecular Weight:
382.88
MDL number:
UNSPSC Code:
12352200
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

ClC1=CC=C(C(NCC2=CC=C(C)C(O)=C2)=C1)NCC3=CC=C(C)C(O)=C3

InChI

1S/C22H23ClN2O2/c1-14-3-5-16(9-21(14)26)12-24-19-8-7-18(23)11-20(19)25-13-17-6-4-15(2)22(27)10-17/h3-11,24-27H,12-13H2,1-2H3

InChI key

WNTAQMQIZGJASL-UHFFFAOYSA-N

Biochem/physiol Actions

Class I glucose transporters (GLUT1-4) inhibitor with greater anti-cancer efficacy than WZB117 (WZB-117) in cultures and in vivo.



DRB18 is a class I glucose transporters (GLUT1-4) inhibitor (glucose uptake IC50 = 2.6/8.8/4.5/0.9 µM using HEK293 GLUT1/2/3/4 transfectants) that exhibits greater anti-cancer efficacy than WZB117 (WZB-117) in cultures (IC50 <10 µM in 51 of 60 cancer cultures tested vs. only 17 from the same 60 cultures when using WZB117) and suppresses A549 xenografts-derived tumor growth in mice in vivo (44% reduction with 10 mg/kg i.p. 3x per wk for 5 wks) by altering the abundance of metabolites in glucose-related pathways.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amanda Westergren Jakobsson et al.
Cancers, 14(13) (2022-07-10)
Neuroblastoma, the most common solid tumor in children, is characterized by amplification of the MYCN proto-oncogene, a high-risk aggressive clinical marker associated with treatment failure. MYCN plays an important role in cell growth, proliferation, metabolism, and chemoresistance. Here, we show
Pratik Shriwas et al.
Cancer & metabolism, 9(1), 14-14 (2021-03-28)
Cancer cells drastically increase the uptake of glucose and glucose metabolism by overexpressing class I glucose transporters (GLUT1-4) to meet their energy and biomass synthesis needs and are very sensitive and vulnerable to glucose deprivation. Although targeting glucose uptake via
Qingzhu Shi et al.
Cancer cell, 40(10), 1207-1222 (2022-09-10)
How glucose metabolism remodels pro-tumor functions of tumor-associated macrophages (TAMs) needs further investigation. Here we show that M2-like TAMs bear the highest individual capacity to take up intratumoral glucose. Their increased glucose uptake fuels hexosamine biosynthetic pathway-dependent O-GlcNAcylation to promote

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service