Skip to Content
Merck
All Photos(1)

Key Documents

SML2384

Sigma-Aldrich

BAY 87-2243

≥98% (HPLC)

Synonym(s):

1-Cyclopropyl-4-[4-[[5-methyl-3-[3-[4-(trifluoromethoxy)phenyl]-1,2,4-oxadiazol-5-yl]-1H-pyrazol-1-yl]methyl]-2-pyridinyl]piperazine, 1-Cyclopropyl-4-{4-[(5-methyl-3-{3-[4-(trifluoromethoxy)phenyl]-1,2,4-oxadiazol-5-yl}-1H-pyrazol-1-yl)methyl]pyridin-2-yl}piperazine

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$118.00
25 MG
$475.00

$118.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$118.00
25 MG
$475.00

About This Item

Empirical Formula (Hill Notation):
C26H26F3N7O2
CAS Number:
Molecular Weight:
525.53
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$118.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

CC1=CC(C2=NC(C3=CC=C(OC(F)(F)F)C=C3)=NO2)=NN1CC4=CC(N5CCN(C6CC6)CC5)=NC=C4

InChI

1S/C26H26F3N7O2/c1-17-14-22(25-31-24(33-38-25)19-2-6-21(7-3-19)37-26(27,28)29)32-36(17)16-18-8-9-30-23(15-18)35-12-10-34(11-13-35)20-4-5-20/h2-3,6-9,14-15,20H,4-5,10-13,16H2,1H3

InChI key

CDJNNOJINJAXPV-UHFFFAOYSA-N

Biochem/physiol Actions

BAY 87-2243 has a therapeutic effect on ferroptosis‐based cancer,[1] hypoxic pancreatic cancer cells.[2]
BAY 87-2243 is an orally active oxidative phosphorylation (OxPhos) inhibitor that potently suppresses hypoxia-induced HIF-1 activation (IC50 = 0.7 nM by HCT-116-based reporter assay; 24 h 1% O2) by selectively targeting mitochondrial complex I (IC50 = 10 nM using mitochondria from PC3; no inhibition of complex III), exhibiting antiproliferation activity in cancer cultures only in the absence of glucose (IC50 ∼3 nM in H460 cultures with 10 mM galactose or lactate, >10 μM with 10 mM glucose). BAY 87-2243 selectively inhibits hypoxia-induced, but not hypoxia-independent, HIF-1α & HIF-2α upregulation and HIF-1 target genes expression in cultures (IC50 ≤10 nM/ADM, ANGPTL4, CA9 mRNA in H460 cells; IC50 = 2 nM/carbonic anhydrase 9 (CA9) protein in HCT116 cells) and effectively suppresses H460 xenograft tumor growth in mice via daily oral admiminstration in vivo (ED50 ∼2 mg/kg).
Highly potent and selective mitochondrial complex I inhibitor that prevents hypoxia-induced HIF-1 activation both in vitro and in vivo.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Matthew J Martin et al.
Oncotarget, 7(52), 86313-86325 (2016-11-20)
Metabolic plasticity is an emerging hallmark of cancer, and increased glycolysis is often observed in transformed cells. Small molecule inhibitors that target driver oncogenes can potentially inhibit the glycolytic pathway. Osimertinib (AZD9291) is a novel EGFR tyrosine kinase inhibitor (TKI)
Richard A Noble et al.
Haematologica, 102(7), 1247-1257 (2017-04-08)
Inhibition of monocarboxylate transporter 1 has been proposed as a therapeutic approach to perturb lactate shuttling in tumor cells that lack monocarboxylate transporter 4. We examined the monocarboxylate transporter 1 inhibitor AZD3965, currently in phase I clinical studies, as a
Gina-Eva Görtz et al.
The Journal of clinical endocrinology and metabolism, 101(12), 4834-4842 (2016-09-10)
In Graves' ophthalmopathy (GO), inflammation with tissue expansion in a closed compartment like the bony orbit and smoking may cause tissue hypoxia. In this study, we investigated whether hypoxia-inducible factor-1 (HIF-1) action impacts on tissue remodeling in GO with the
Linda Helbig et al.
Radiation oncology (London, England), 9, 207-207 (2014-09-23)
The transcription factor hypoxia-inducible factor-1 (HIF-1) pathway plays an important role in tumor response to cytotoxic treatments. We investigated the effects of a novel small molecule inhibitor of mitochondrial complex I and hypoxia-induced HIF-1 activity BAY-87-2243, on tumor microenvironment and
Emerging strategies of cancer therapy based on ferroptosis
Shen z, et al.
Advanced Materials, 30(12), 1704007-1704007 (2018)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service