Skip to Content
Merck
All Photos(1)

Key Documents

SML1779

Sigma-Aldrich

Nigericin sodium salt

from Streptomyces hygroscopicus, ≥98% (HPLC), solution, polyether ionophore

Synonym(s):

3B2-6379, Antibiotic K178, Antibiotic X464, Azalomycin M, HE331800, Helexin C, Polyetherin A, sodium;2-[(3S,6R)-6-[[(5R,6R,7R,9R)-2-[5-[(3S,5R)-5-[(2S,3S,5R,6R)-6-hydroxy-6-(hydroxymethyl)-3,5-dimethyloxan-2-yl]-3-methyloxolan-2-yl]-5-methyloxolan-2-yl]-7-methoxy-2,4,6-trimethyl-1,10-dioxaspiro[4.5]decan-9-yl]methyl]-3-methyloxan-2-yl]propanoate

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
$189.00

$189.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
$189.00

About This Item

Empirical Formula (Hill Notation):
C40H67NaO11
CAS Number:
Molecular Weight:
746.94
UNSPSC Code:
12352200
NACRES:
NA.77

$189.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Product Name

Nigericin sodium salt Ready Made Solution, 5 mg/mL (DMSO:ethanol 1:1)

biological source

Streptomyces hygroscopicus

Quality Level

form

solution

concentration

5 mg/mL (DMSO:ethanol 1:1)

antibiotic activity spectrum

Gram-positive bacteria

Mode of action

cell membrane | interferes

shipped in

ambient

storage temp.

2-8°C

SMILES string

[Na+].[O-]C(=O)C(C1O[C@H](CC[C@@H]1C)C[C@H]2O[C@]3(OC(CC3C)(C4OC(CC4)(C5O[C@H](C[C@@H]5C)[C@H]6O[C@@]([C@@H](C[C@@H]6C)C)(O)CO)C)C)[C@@H]([C@@H](C2)OC)C)C

InChI

1S/C40H68O11.Na/c1-21-11-12-28(46-33(21)26(6)36(42)43)17-29-18-30(45-10)27(7)40(48-29)25(5)19-38(9,51-40)32-13-14-37(8,49-32)35-23(3)16-31(47-35)34-22(2)15-24(4)39(44,20-41)50-34;/h21-35,41,44H,11-20H2,1-10H3,(H,42,43);/q;+1/p-1/t21-,22-,23-,24+,25?,26?,27+,28+,29+,30+,31+,32?,33?,34-,35?,37?,38?,39-,40+;/m0./s1

InChI key

MOYOTUKECQMGHE-KKCUGXASSA-M

Biochem/physiol Actions

Nigericin is a polyether ionophore which catalyzes the electroneutral exchange of alkali metal (K+) for H+ (antiport).
Nigericin is a polyether ionophore which catalyzes the electroneutral exchange of alkali metal (K+) for H+ (antiport). Nigericin transports monovalent cations across membranes with the following specificity: K+ >Rb+ >Cs+ >>Na+ and thus, disrupts membrane potential and stimulates ATPase activity in mitochondria.
Nigericin kills bacteria by facilitating the diffusion of ions across membranes.
Low concentration (0.5 μM) of Nigericin rapidly decreases pHi, causing stimulation of PG production 1.5- to 2-fold in cerebral microvascular endothelial cells and arresting of DNA synthesis in Erlich acites carcinoma cells. Treatment of Hela cells, after entry of poliovirus, with nigericin, prevents the inhibition of host protein synthesis by poliovirus. Nigericin is also widely used in studies of the consequences of changes in membrane potential in variable systems.

Preparation Note

Nigericin sodium salt ready made solution is provided at 6.7 mM concentration. The solution can be further diluted with a desired solvent to a working concentration (0.5-10 μM).
This product is a 5 mg/mL solution prepared in a 1:1 mixture of DMSO and Ethanol.

Storage and Stability

The product can be stored at 2-8°C and is released with a 4 years suggested retest schedule. The solution is quite stable refrigerated and freezing is not necessary.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

78.8 °F

Flash Point(C)

26 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Irurzun et al.
Journal of virology, 69(12), 7453-7460 (1995-12-01)
Addition of monensin or nigericin after poliovirus entry into HeLa cells prevents the inhibition of host protein synthesis by poliovirus. The infected cells continue to synthesize cellular proteins at control levels for at least 8 h after infection in the
Moran Morelli et al.
Scientific reports, 14(1), 5797-5797 (2024-03-10)
Enterotoxins are a type of toxins that primarily affect the intestines. Understanding their harmful effects is essential for food safety and medical research. Current methods lack high-throughput, robust, and translatable models capable of characterizing toxin-specific epithelial damage. Pressing concerns regarding
Hachiro Konaka et al.
The EMBO journal, 42(20), e112573-e112573 (2023-09-04)
Mitochondrial DNA (mtDNA) leakage into the cytoplasm can occur when cells are exposed to noxious stimuli. Specific sensors recognize cytoplasmic mtDNA to promote cytokine production. Cytoplasmic mtDNA can also be secreted extracellularly, leading to sterile inflammation. However, the mode of
L B Margolis et al.
Proceedings of the National Academy of Sciences of the United States of America, 86(17), 6626-6629 (1989-09-01)
Acidification of the cytoplasm of Ehrlich ascites carcinoma cells to pH 6.3 arrests DNA synthesis in these cells. Such an effect can be achieved by incubating the cells at pH 6.2 or by adding low concentrations of the K+/H+ antiporter
Shigetoshi Yokoyama et al.
Cell death discovery, 7(1), 12-12 (2021-01-17)
Non-canonical inflammasome activation that recognizes intracellular lipopolysaccharide (LPS) causes pyroptosis, the inflammatory death of innate immune cells. The role of pyroptosis in innate immune cells is to rapidly eliminate pathogen-infected cells and limit the replication niche in the host body.

Questions

  1. How stable is the antibiotic in solution, how long can I store it for? Can I freeze a few aliquots in -80 deg C, or will it cause the chemical to denature?

    1 answer
    1. This product is a 5 mg/mL solution prepared in a 1:1 mixture of DMSO and Ethanol. The product is stored at 2-8°C and is released with a 4 years suggested retest schedule. The solution is quite stable refrigerated and freezing is not necessary. However, the pure powder product (N7143) is stable at -20°C for at least 3 months when dissolved in DMSO or Ethanol. Storage at less than -20°C has not been evaluated. Please see the link below for a sample Certificate of Analysis which includes the recommended retest date:
      https://www.sigmaaldrich.com/certificates/sapfs/PROD/sap/certificate_pdfs/COFA/Q14/SML1779-1ML0000225533.pdf

      Please see the link below to review the product datasheet for the powdered Nigericin, N7143:
      https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/379/493/n7143pis.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service