Skip to Content
Merck
All Photos(1)

Key Documents

SML1735

Sigma-Aldrich

PFM01

≥98% (HPLC)

Synonym(s):

(5Z)-5-[(4-Hydroxyphenyl)methylene]-3-(2-methylpropyl)-2-thioxo-4-thiazolidinone, 5-(4-Hydroxybenzylidene)-3-isobutyl-2-thioxothiazolidin-4-one

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$143.00
25 MG
$583.00

$143.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
5 MG
$143.00
25 MG
$583.00

About This Item

Empirical Formula (Hill Notation):
C14H15NO2S2
CAS Number:
Molecular Weight:
293.40
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$143.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

OC1=CC=C(/C=C2SC(N(CC(C)C)C\2=O)=S)C=C1

InChI

1S/C14H15NO2S2/c1-9(2)8-15-13(17)12(19-14(15)18)7-10-3-5-11(16)6-4-10/h3-7,9,16H,8H2,1-2H3/b12-7-

InChI key

GPURHDUTZUYAFI-GHXNOFRVSA-N

Biochem/physiol Actions

PFM01 is a cell-permeable N-alkylated Mirin (Sigma Cat. No. 475954) derivative that selectively inhibits against MRE11 endo-, but not exo-, nuclease activity. PFM01 targets MRE11 at a site near the dimer interface, distinct from that occupied by Mirin and PFM39 to allow disruption of the ssDNA-binding groove and selective inhibition against MRE11 endo-, but not exo-, nuclease activity. While both endonuclease and exonuclease activities are required for MRE11-mediated homologous recombination (HR) repair, only FM01 (100 μM), but not the exonuclease inhibitors Mirin (500 μM) and PFM39 (100 μM), rescues G2-phase double-strand break (DSB) repair defect in HR protein BRCA2-deficient HSC62-hTERT fibroblasts following ionizing irradiation (IR) by blocking HR initiation and thereby allowing non-homologous end joining (NHEJ) to proceed.
PFM01 is a cell-permeable N-alkylated Mirin derivative that selectively inhibits against MRE11 endo-, but not exo-, nuclease activity.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lea Völkening et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 34(2), 2812-2820 (2020-01-08)
The Mre11A/RAD50/NBN complex (MRN) is an essential regulator of the cellular damage response after DNA double-strand breaks (DSBs). More recent work has indicated that MRN may also impact on the duration of mitosis. We show here that RAD50-deficient fibroblasts exhibit
Sarah R Hengel et al.
Cell chemical biology, 24(9), 1101-1119 (2017-09-25)
To maintain stable genomes and to avoid cancer and aging, cells need to repair a multitude of deleterious DNA lesions, which arise constantly in every cell. Processes that support genome integrity in normal cells, however, allow cancer cells to develop
Donna R Whelan et al.
Proceedings of the National Academy of Sciences of the United States of America, 118(11) (2021-03-13)
Homologous recombination (HR) is a major pathway for repair of DNA double-strand breaks (DSBs). The initial step that drives the HR process is resection of DNA at the DSB, during which a multitude of nucleases, mediators, and signaling proteins accumulates

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service