Skip to Content
Merck
All Photos(1)

Key Documents

SML3088

Sigma-Aldrich

MRK-016

≥98% (HPLC)

Synonym(s):

3-(1,1-Dimethylethyl)-7-(5-methyl-3-isoxazolyl)-2-[(1-methyl-1H-1,2,4-triazol-5-yl)methoxy]-pyrazolo[1,5-d][1,2,4]triazine, 3-tert-Butyl-7-(5-methylisoxazol-3-yl)-2-(1-methyl-1H-1,2,4-triazol-5-ylmethoxy)-pyrazolo[1,5-d][1,2,4]triazine, MRK 016, MRK016

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$183.00
25 MG
$584.00

$183.00


Estimated to ship onMay 25, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$183.00
25 MG
$584.00

About This Item

Empirical Formula (Hill Notation):
C17H20N8O2
CAS Number:
Molecular Weight:
368.39
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$183.00


Estimated to ship onMay 25, 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

CC(C)(C1=C2C=NN=C(N2N=C1OCC3=NC=NN3C)C4=NOC(C)=C4)C

InChI

1S/C17H20N8O2/c1-10-6-11(23-27-10)15-21-19-7-12-14(17(2,3)4)16(22-25(12)15)26-8-13-18-9-20-24(13)5/h6-7,9H,8H2,1-5H3

InChI key

QYSYOGCIDRANAR-UHFFFAOYSA-N

Biochem/physiol Actions

MRK-016 is an orally active, benzodiazepine site-targeting, alpha5-selective GABAA inverse agonist (Emax = -55%/α5, -16%/α1, -9%/α3, +6%/α2 of a EC20 GABA response using respective recombinant human αβ3γ2) with in vivo cognition-enhancing properties (0.3-3 mg/kg p.o. in rat Morris water maze test).
Orally active, benzodiazepine site-targeting, alpha5 (α5) subtype-selective GABAA inverse agonist with cognition-enhancing properties in vivo.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiumin Chen et al.
Neuropharmacology, 125, 243-253 (2017-08-02)
α5-containing GABAARs are potential therapeutic targets for clinical conditions including age-related dementia, stroke, schizophrenia, Down syndrome, anaesthetic-induced amnesia, anxiety and pain. α5-containing GABAARs are expressed in layer 5 cortical neurons and hippocampal pyramidal neurons where they mediate both tonic currents

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service