Skip to Content
Merck
All Photos(1)

Documents

SML2170

Sigma-Aldrich

HX531

≥98% (HPLC)

Synonym(s):

4-(5H-2,3-(2,5-dimethyl-2,5-hexano)-5-methyl- 8-nitrodibenzo[b,e][1,4]diazepin-11-yl)benzoic acid, 4-(7,8,9,10-Tetrahydro-5,7,7,10.10-pentamethyl-2-nitro-5H-benzo[b]naphtho[2,3-e][1,4]-diazepin-12-yl)-benzoic acid, HX 531, HX-531

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H29N3O4
CAS Number:
Molecular Weight:
483.56
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

yellow to orange

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

CN(C1=CC2=C(C(C)(CCC2(C)C)C)C=C13)C(C=C4)=C(N=C3C5=CC=C(C=C5)C(O)=O)C=C4[N+]([O-])=O

InChI

1S/C29H29N3O4/c1-28(2)12-13-29(3,4)22-16-25-20(15-21(22)28)26(17-6-8-18(9-7-17)27(33)34)30-23-14-19(32(35)36)10-11-24(23)31(25)5/h6-11,14-16H,12-13H2,1-5H3,(H,33,34)

InChI key

SXKPGYKPQPYJER-UHFFFAOYSA-N

Biochem/physiol Actions

HX-531 is a potent antagonist of the retinoid X receptor (RXR) with an IC50 value of 18 nM. HX531 has been used in many studies of the various effects of retinoids and RXR activity in diseases such as diabetes, Parkinson′s disease, often as a control for agonists such as Bexarotene. It was recently shown to promote white adipogenic differentiation and inhibit both basal and Bexarotene-induced brown adipogenic differentiation.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service