Skip to Content
Merck
All Photos(1)

Key Documents

SML2855

Sigma-Aldrich

LY2603618

≥98% (HPLC)

Synonym(s):

(S)-1-[5-Bromo-4-methyl-2-(morpholin-2-ylmethoxy)phenyl]-3-(5-methylpyrazin-2-yl)urea, 1-[5-Bromo-4-methyl-2-(S)-(morpholin-2-ylmethoxy)phenyl]-3-(5-methylpyrazin-2-yl)urea, IC-83, LCI-1, LY 2603618, LY-2603618, Rabusertib

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$118.00
25 MG
$327.00

$118.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$118.00
25 MG
$327.00

About This Item

Empirical Formula (Hill Notation):
C18H22BrN5O3
CAS Number:
Molecular Weight:
436.30
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$118.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

CC1=C(Br)C=C(NC(NC2=CN=C(C)C=N2)=O)C(OC[C@H]3OCCNC3)=C1

InChI

1S/C18H22BrN5O3/c1-11-5-16(27-10-13-8-20-3-4-26-13)15(6-14(11)19)23-18(25)24-17-9-21-12(2)7-22-17/h5-7,9,13,20H,3-4,8,10H2,1-2H3,(H2,22,23,24,25)/t13-/m0/s1

InChI key

SYYBDNPGDKKJDU-ZDUSSCGKSA-N

Biochem/physiol Actions

LY2603618 (LCI-1) is a potent and selective checkpoint kinase 1 (Chk1) inhibitor (IC50 = 7 nM) that produces a cellular phenotype identical to that reported upon Chk1 depletion by RNAi. LY2603618 prevents doxorubicin-induced Chk1 autophosphorylation (IC50 = 52 nM; HeLa), allowing cells to traverse the G2/M checkpoint and proceed into mitosis with unresolved replicated chromosomes. LY2603618 renders mutant p53, but not wild-type, HT-29 colon cancer cells more sensitive to gemcitabine both in vitro and in mice in vivo.
Potent and selective checkpoint kinase 1 (Chk1) inhibitor that renders mutant p53 cancer cells more sensitive to DNA-damaging agents in vitro and in vivo.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Characterization and preclinical development of LCI-1, a selective and potent Chk1 inhibitor in phase I clinical trials
Marshall M, Barda D, Barnard D, et al.
Molecular Cancer Therapeutics, 8, B248-B248 (2009)
Constance King et al.
Investigational new drugs, 32(2), 213-226 (2013-10-12)
Interference with DNA damage checkpoints has been demonstrated preclinically to be a highly effective means of increasing the cytotoxicity of a number of DNA-damaging cancer therapies. Cell cycle arrest at these checkpoints protects injured cells from apoptotic cell death until
Darlene Barnard et al.
Investigational new drugs, 34(1), 49-60 (2015-11-28)
Pharmacological inhibition of CHK1 in the absence of p53 functionality leads to abrogation of the S and G2/M DNA damage checkpoints. We report the preclinical therapeutic activity of LY2603618 (CHK1 inhibitor) at inhibiting CHK1 activation by gemcitabine and enhancing in
Anne M van Harten et al.
Oncogenesis, 8(7), 38-38 (2019-06-19)
Head and neck squamous cell carcinomas (HNSCCs) coincide with poor survival rates. The lack of driver oncogenes complicates the development of targeted treatments for HNSCC. Here, we follow-up on two previous genome-wide RNA and microRNA interference screens in HNSCC to
Jianyun Zhao et al.
Oncotarget, 7(23), 34785-34799 (2016-05-12)
Resistance to standard chemotherapy agents remains a major obstacle for improving treatment outcomes for acute myeloid leukemia (AML). The Bcl-2-selective inhibitor ABT-199 has demonstrated encouraging preclinical results, drug resistance remains a concern. Mcl-1 has been demonstrated to contribute to ABT-199

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service