Skip to Content
Merck
All Photos(1)

Key Documents

14340

Sigma-Aldrich

(+)-Bicuculline

≥97.0% (TLC)

Synonym(s):

Bucuculline

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$82.60
100 MG
$127.00
500 MG
$507.00

$82.60


Estimated to ship onMay 18, 2025


Request a Bulk Order

Select a Size

Change View
25 MG
$82.60
100 MG
$127.00
500 MG
$507.00

About This Item

Empirical Formula (Hill Notation):
C20H17NO6
CAS Number:
Molecular Weight:
367.35
Beilstein:
98786
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$82.60


Estimated to ship onMay 18, 2025


Request a Bulk Order

Quality Level

Assay

≥97.0% (TLC)

form

powder

optical activity

[α]20/D +126±6°, c = 1% in chloroform

mp

193-197 °C

SMILES string

[H][C@]1(OC(=O)c2c3OCOc3ccc12)[C@@]4([H])N(C)CCc5cc6OCOc6cc45

InChI

1S/C20H17NO6/c1-21-5-4-10-6-14-15(25-8-24-14)7-12(10)17(21)18-11-2-3-13-19(26-9-23-13)16(11)20(22)27-18/h2-3,6-7,17-18H,4-5,8-9H2,1H3/t17-,18+/m0/s1

InChI key

IYGYMKDQCDOMRE-ZWKOTPCHSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Bicuculline is a convulsant alkaloid.[1] It was originally isolated from the plant Dicentra cucullaria. [2]

Application

Bicuculline has been used:
  • as a compound to compare pharmacodynamics and network activity profiles of conolidine/cannabidiol[3]
  • to study the effects of chronic caffeine administration on the function of GABAA receptor[4]
  • to isolate N-methyl-D-aspartate receptor (NMDAR)-specific evoked and miniature excitatory postsynaptic currents (eEPSCs and mEPSCs) in neurons of rats[5]

Biochem/physiol Actions

(+)-Bicuculline acts as a competitive inhibitor of GABA liganding binding to the receptor.[6]
(+)-Bicuculline is a GABAA receptor antagonist.

Features and Benefits

This compound is featured on the GABAC Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Discovering the pharmacodynamics of conolidine and cannabidiol using a cultured neuronal network based workflow
Mendis GDC, et al.
Scientific reports, 9(1), 121-121 (2019)
Herbal products and GABA receptors
Johnston GAR, et al.
Encyclopedia of Neuroscience, 169(2), 1095-1101 (2009)
Evidence that bicuculline and picrotoxin act at separate sites to antagonize gamma-aminobutyric acid in rat cuneate nucleus.
M A Simmonds
Neuropharmacology, 19(1), 39-45 (1980-01-01)
Prolonged ketamine exposure induces increased activity of the GluN2B-containing N-methyl-D-aspartate receptor in the anterior cingulate cortex of neonatal rats
Kokane SS, et al.
Neurotoxicology and Teratology, 63(2), 1-8 (2017)
S Ueno et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 17(2), 625-634 (1997-01-15)
Anesthetic drugs are known to interact with GABAA receptors, both to potentiate the effects of low concentrations of GABA and to directly gate open the ion channel in the absence of GABA; however, the site(s) involved in direct gating by

Articles

Yeast is one of the most important microorganisms known and utilised by mankind. Ancient Middle Eastern civilisations used the organism to bake bread and to produce mead, beer and wine.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service