Skip to Content
Merck
All Photos(1)

Key Documents

SML2593

Sigma-Aldrich

MS0021570

≥98% (HPLC)

Synonym(s):

5-(Benzylsulfanyl)-N-methyl-1,3,4-thiadiazol-2-amine, 5-(Benzylthio)-N-methyl-1,3,4-thiadiazol-2-amine, MS 21570, MS0021570_1, N-Methyl-5-[(phenylmethyl)thio]-1,3,4-thiazadiazol-2-amine

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$122.00
25 MG
$493.00

$122.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$122.00
25 MG
$493.00

About This Item

Empirical Formula (Hill Notation):
C10H11N3S2
CAS Number:
Molecular Weight:
237.34
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$122.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

[s]1c(nnc1NC)SCc2ccccc2

InChI

1S/C10H11N3S2/c1-11-9-12-13-10(15-9)14-7-8-5-3-2-4-6-8/h2-6H,7H2,1H3,(H,11,12)

InChI key

KFLNRVSOQJTXDG-UHFFFAOYSA-N

Biochem/physiol Actions

GPR171 antagonist
MS0021570 is a potent antagonist of GPR171, a G protein-coupled receptor of BigLEN (b-LEN), which is a neuropeptide derived from proSAAS and involved in regulation of feeding. MS0021570 had a IC50 value of 220 nM. It was shown to block inceases in food intake and to reduce anxiety-like behavior and fear conditioning in mice.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service