Skip to Content
Merck
All Photos(1)

Key Documents

SML2472

Sigma-Aldrich

UPGL00004

≥98% (HPLC)

Synonym(s):

2-Phenyl-N-(5-(4-((5-(2-phenylacetamido)-1,3,4-thiadiazol-2-yl)amino)piperidin-1-yl)-1,3,4-thiadiazol-2-yl)acetamide, N-[5-[4-[[5-[(2-Phenylacetyl)amino]-1,3,4-thiadiazol-2-yl]amino]-1-piperidinyl]-1,3,4-thiadiazol-2-yl]benzeneacetamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$109.00
25 MG
$351.00

$109.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$109.00
25 MG
$351.00

About This Item

Empirical Formula (Hill Notation):
C25H26N8O2S2
CAS Number:
Molecular Weight:
534.66
UNSPSC Code:
12352200
NACRES:
NA.77

$109.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to pink

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(CC1=CC=CC=C1)NC(S2)=NN=C2N(CC3)CCC3NC4=NN=C(NC(CC5=CC=CC=C5)=O)S4

InChI

1S/C25H26N8O2S2/c34-20(15-17-7-3-1-4-8-17)27-23-30-29-22(36-23)26-19-11-13-33(14-12-19)25-32-31-24(37-25)28-21(35)16-18-9-5-2-6-10-18/h1-10,19H,11-16H2,(H,26,29)(H,27,30,34)(H,28,31,35)

InChI key

MRYCNTHLPRENBA-UHFFFAOYSA-N

Biochem/physiol Actions

UPGL00004, a BPTES analog, is an orally available, potent and selective inhibitor of glutaminase C (GAC) that potently inhibits the growth of triplenegative breast cancer cells. UPGL00004 directly binds to the allosteric site at GAC.
orally available, potent and selective inhibitor of glutaminase C (GAC)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service