Skip to Content
Merck
All Photos(1)

Key Documents

SML1802

Sigma-Aldrich

MS0015203

≥98% (HPLC)

Synonym(s):

5-[(2-Methyl-1-oxo-2-propen-1-yl)amino]-1,3-benzenedicarboxylic acid

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$122.00

$122.00


Estimated to ship onMay 26, 2025



Select a Size

Change View
5 MG
$122.00

About This Item

Empirical Formula (Hill Notation):
C12H11NO5
CAS Number:
Molecular Weight:
249.22
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$122.00


Estimated to ship onMay 26, 2025


Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(O)C1=CC(C(O)=O)=CC(NC(C(C)=C)=O)=C1

InChI

1S/C12H11NO5/c1-6(2)10(14)13-9-4-7(11(15)16)3-8(5-9)12(17)18/h3-5H,1H2,2H3,(H,13,14)(H,15,16)(H,17,18)

InChI key

NVOBVSWSDYFEMR-UHFFFAOYSA-N

Biochem/physiol Actions

MS0015203 is a potent and selective agonist of the neuropeptide receptor GPR171.
MS0015203 is a potent and selective agonist of the neuropeptide receptor GPR171. MS0015203 increases food intake and body weight in mice.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service