Skip to Content
Merck
All Photos(1)

Key Documents

SML0136

Sigma-Aldrich

TGN-020

≥98% (HPLC)

Synonym(s):

2-(Nicotinamide)-1,3,4-thiadiazole, N-(1,3,4-Thiadiazol-2-yl)-3-pyridinecarboxamide, N-(1,3,4-Thiadiazol-2-yl)pyridine-3-carboxamide, N-(1,3,4-Thiadiazolyl)-m-nicotinamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$186.00
25 MG
$738.00

$186.00


Estimated to ship onMay 28, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$186.00
25 MG
$738.00

About This Item

Empirical Formula (Hill Notation):
C8H6N4OS
CAS Number:
Molecular Weight:
206.22
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$186.00


Estimated to ship onMay 28, 2025


Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to tan

solubility

DMSO: ≥8 mg/mL

storage temp.

room temp

SMILES string

O=C(Nc1nncs1)c2cccnc2

InChI

1S/C8H6N4OS/c13-7(6-2-1-3-9-4-6)11-8-12-10-5-14-8/h1-5H,(H,11,12,13)

InChI key

AGEGZHOPKZFKBP-UHFFFAOYSA-N

Application

Beas2B human lung epithelial cells were treated with TGN-020 to study the effect on cigarette smoke extracts-induced CCN1 secretion.[1]

Biochem/physiol Actions

Aquaporin 4 (AQP4) inhibitor
TGN-020 is an inhibitor of Aquaporin 4 (AQP4), the most abundant water channel in brain. Aquaporins (AQPs) are water channels required for maintaining fluid homeostasis and enabling water movement across barrier membranes, but can enhance pathological cellular volume changes and cause edema in injury states. Pretreatment with the AQP4 inhibitor TGN-020 significantly reduced the volume of brain edema associated with ischemic injury in a mouse model of focal cerebral ischemia.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hyung-Geun Moon et al.
PloS one, 8(7), e68199-e68199 (2013-07-23)
Inflammation involves in many cigarette smoke (CS) related diseases including the chronic obstructive pulmonary disease (COPD). Lung epithelial cell released IL-8 plays a crucial role in CS induced lung inflammation. CS and cigarette smoke extracts (CSE) both induce IL-8 secretion
Hiromu Monai et al.
Proceedings of the National Academy of Sciences of the United States of America, 116(22), 11010-11019 (2019-05-18)
Spontaneous waves of cortical spreading depolarization (CSD) are induced in the setting of acute focal ischemia. CSD is linked to a sharp increase of extracellular K+ that induces a long-lasting suppression of neural activity. Furthermore, CSD induces secondary irreversible damage
Jian Li et al.
Brain research bulletin, 170, 162-173 (2021-02-17)
The extracellular signal-regulated kinase (ERK) pathway has been reported to play a pivotal role in mediating spinal cord injury (SCI) progression. The present study aimed to investigate the effects of phosphorylated ERK1/2 (p-ERK1/2) inhibition on SCI-induced astrocyte activation and inflammation
Dervis Dasdelen et al.
Mini reviews in medicinal chemistry, 20(6), 498-512 (2019-10-28)
In the literature screening, aquaporins were found in the cerebral structures including the pia mater, choroid plexus, ependyma, piriform cortex, hippocampus, dorsal thalamus, supraoptic and suprachiasmatic nuclei, white matter and subcortical organ. Among these, the most common are AQP1, AQP4
Cheng Guo et al.
Inflammation, 42(4), 1401-1412 (2019-04-05)
Acute lung injury (ALI) is a syndrome characterized by damage to the alveolar-capillary wall, pulmonary edema and recruitment of inflammatory cells. Previous studies have indicated that aquaporin 4 (AQP4) plays a key role in brain edema formation and resolution. However

Questions

  1. We purchased a quantity of TGN-020 earlier this year but have had exceptional difficulty finding an effective solvent for this compound (tried DCM and H2O). Does Sigma-Aldrich have data on compatible solvents? Primary lit hasn't been helpful. Thanks!

    1 answer
    1. The solubility specification for this product is >/= 8mg/mL in DMSO. The rate may be higher and is lot-specific, however, the minimum concentration in DMSO is 8 mg/mL. A brief literature search shows that HCl, Ethanol, and Dimethylformamide have also been used to dissolve this compound.
      Please see the sample Certificate of Analysis for example:
      https://www.sigmaaldrich.com/certificates/sapfs/PROD/sap/certificate_pdfs/COFA/Q14/SML0136-5MG0000258843.pdf

      If solubility issues continue, we kindly ask you to navigate to the link https://www.sigmaaldrich.com/techservice, and click on "Report Product Issues" under the Products Section with all the required information so that a member of our team can reach out to you to assist further. Thank you.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service