Skip to Content
Merck
All Photos(1)

Key Documents

P1174

Sigma-Aldrich

Microcapillary tube, calibrated

capacity 100 μL

Synonym(s):

Calibrated Capillary Tube, Capillary Tube, Microcapillary Tube

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
$234.00

$234.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025



Select a Size

Change View
1 PKG
$234.00

About This Item

UNSPSC Code:
41121500
NACRES:
NB.01

$234.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


material

glass

packaging

pack of 250 ea

manufacturer/tradename

Drummond Scientific 2-000-100

measuring range

accuracy: ±0.5%

capacity

100 μL

color

color-coded blue

Looking for similar products? Visit Product Comparison Guide

General description

  • Drummond Calibrated Micropipettes provide accuracy and convenience for a wide variety of liquid transfer applications.
  • Made of high quality glass.
  • Fire polished micropipettes are imprinted with volume and a color-code bar for easy identification.
  • Calibrated "To Deliver."
High quality Drummond micropipettes are fire polished, imprinted with volume and a color-code bar for easy identification. Calibrated "To Deliver."

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–2 of 2 Questions  
  1. Can the product be used for serum? Also, does it contain EDTA or heparin?

    1 answer
    1. The Drummond Scientific 2-000-100 calibrated pipets, made from borosilicate glass, offer a ±0.5% accuracy and are suitable for various liquid sample handling tasks. Each vial contains 250 individual pipets and one 15″ aspirator tube. These pipets are available in sizes ranging from 1-200 microliters, with heparinized options for blood transfer work. The P1174 Microcapillary tube, calibrated, is not heparinized. For blood transfer work, a heparinized 44.7 microliter pipet (2-000-044-H) is also available.

      Helpful?

  2. Could you provide information about the dimensions of the Microcapillary tube that is calibrated and has a capacity of 100 microliters? Specifically, I'm interested in knowing the length and depth of this product.

    1 answer
    1. This calibrated pipette has an outer diameter/inner diameter of 1.689/1.3030 mm. All of our calibrated pipettes are 5" (127mm) in length.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service