Key Documents
P4925
Sigma-Aldrich
Pipette filler for calibrated microcapillary pipettes
dispenses up to 100 μL
Select a Size
1 EA
$196.00
$196.00
PRICE WITHOUT NATIONAL TAXES
Estimated to ship onJune 01, 2025
Select a Size
Change View
1 EA
$196.00
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
$196.00
PRICE WITHOUT NATIONAL TAXES
Estimated to ship onJune 01, 2025
material
aluminum (Anodized)
blue
packaging
pack of 1 ea
manufacturer/tradename
Drummond Scientific 3-000-752
capacity
1-100 μL
General description
Anodized aluminum body and screw-operated piston aspirates and dispenses up to 100 μL. Tapered silicone rubber neck holds all sizes of capillaries securely.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Liyang Zhang et al.
Genome research, 28(1), 111-121 (2017-12-03)
The DNA-binding interfaces of the androgen (AR) and glucocorticoid (GR) receptors are virtually identical, yet these transcription factors share only about a third of their genomic binding sites and regulate similarly distinct sets of target genes. To address this paradox
Julio C Sanchez et al.
Biochimica et biophysica acta. Gene regulatory mechanisms, 1863(8), 194566-194566 (2020-05-08)
The ATP-dependent BAF chromatin remodeling complex plays a critical role in gene regulation by modulating chromatin architecture, and is frequently mutated in cancer. Indeed, subunits of the BAF complex are found to be mutated in >20% of human tumors. The