Skip to Content
Merck
All Photos(4)

Key Documents

N7143

Sigma-Aldrich

Nigericin sodium salt

≥98% (TLC)

Synonym(s):

Antibiotic K178, Antibiotic X464, Azalomycin M, Helexin C, Polyetherin A

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$166.00
10 MG
$298.00
50 MG
$1,050.00

$166.00


Estimated to ship onMarch 22, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$166.00
10 MG
$298.00
50 MG
$1,050.00

About This Item

Empirical Formula (Hill Notation):
C40H67NaO11
CAS Number:
Molecular Weight:
746.94
Beilstein:
3892398
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$166.00


Estimated to ship onMarch 22, 2025


Request a Bulk Order

biological source

Streptomyces hygroscopicus

Quality Level

Assay

≥98% (TLC)

form

powder

antibiotic activity spectrum

Gram-positive bacteria

Mode of action

cell membrane | interferes

storage temp.

2-8°C

SMILES string

[Na+].[H][C@@]1(CC[C@H](C)[C@@]([H])(O1)[C@@H](C)C([O-])=O)C[C@]2([H])C[C@@H](OC)[C@@H](C)[C@]3(O2)O[C@@](C)(C[C@H]3C)[C@@]4([H])CC[C@](C)(O4)[C@]5([H])O[C@]([H])(C[C@@H]5C)[C@@]6([H])O[C@@](O)(CO)[C@H](C)C[C@@H]6C

InChI

1S/C40H68O11.Na/c1-21-11-12-28(46-33(21)26(6)36(42)43)17-29-18-30(45-10)27(7)40(48-29)25(5)19-38(9,51-40)32-13-14-37(8,49-32)35-23(3)16-31(47-35)34-22(2)15-24(4)39(44,20-41)50-34;/h21-35,41,44H,11-20H2,1-10H3,(H,42,43);/q;+1/p-1/t21-,22-,23-,24+,25+,26+,27+,28+,29+,30+,31+,32+,33+,34-,35+,37-,38-,39-,40+;/m0./s1

InChI key

MOYOTUKECQMGHE-PDEFJWSRSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: polyether
Nigericin, a polyether ionophore, serves as a probe of ion transport across mitochondrial membranes.[1] It catalyzes the overall electroneutral exchange of K+ for H+(1) and acts as a potassium efflux activator. It may enhance bactericidal action via inflammasome-independent mechanisms specific to NLRP3 (NOD-, LRR- and pyrin domain-containing protein 3).[2] Nigericin is a linear molecule with rings of heterocyclic oxygen together with a hydroxyl group. In the membrane, the molecule cyclizes to form a structure like valinomycin. Nigericin forms a neutral complex while losing a proton when it binds a cation which can diffuse across the membrane as a mobile carrier. Nigericin in protonated noncomplexed form is also considered mobile.

Application

Nigericin sodium salt has been used to induce NLRP3 inflammasome activation in human macrophages.[3] It has also been used as a bacterial toxin to trigger IL-1β secretion monocytes.[4]

Biochem/physiol Actions

Polyether ionophore that disrupts membrane potential and stimulates ATPase activity in mitochondria. Ion selectivity is K+> Rb+≥ Cs+>> Na+.

Other Notes

As ion-selective electrode - an efficient K+-carrier.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mohamed Wehbe et al.
International journal of nanomedicine, 12, 4129-4146 (2017-06-16)
Copper diethyldithiocarbamate (Cu(DDC)
Adesola C Olatunde et al.
PloS one, 13(5), e0196591-e0196591 (2018-05-11)
Dendritic cells (DC) are sentinels of the immune system, alerting and enlisting T cells to clear pathogenic threats. As such, numerous studies have demonstrated their effective uptake and proteolytic activities coupled with antigen processing and presentation functions. Yet, less is
H Parfenova et al.
The American journal of physiology, 277(4 Pt 1), C728-C738 (1999-10-12)
Nigericin decreases intracellular pH (pH(i)) and stimulates prostanoid (PG) synthesis in endothelial cells from cerebral microvessels of newborn pigs. Nigericin-induced PG production was abolished by protein tyrosine kinase (PTK) inhibitors and amplified by phorbol 12-myristate 13-acetate (PMA) or protein tyrosine
Nardhy Gomez-Lopez et al.
Biology of reproduction, 100(5), 1306-1318 (2019-01-01)
Sterile intra-amniotic inflammation is commonly observed in patients with spontaneous preterm labor, a syndrome that commonly precedes preterm birth, the leading cause of perinatal morbidity and mortality worldwide. However, the mechanisms leading to sterile intra-amniotic inflammation are poorly understood and
Leonid Tarassishin et al.
PloS one, 9(7), e103432-e103432 (2014-07-24)
Glioblastoma is the most frequent and malignant form of primary brain tumor with grave prognosis. Mounting evidence supports that chronic inflammation (such as chronic overactivation of IL-1 system) is a crucial event in carcinogenesis and tumor progression. IL-1 also is

Questions

  1. Has nigericin been tested or approved for intraperitoneal (i.p.) injection in mice?also advice on the safety of nigericin injection into mice and recommended solvents for its use.

    1 answer
    1. The product N7143 is intended for Research Use Only and does not have approvals from any agency for i.p. injections. Nigericin is soluble in DMSO up to 50 mg/mL and in ethanol up to 20 mg/mL, with stock solutions able to be stored for up to 3 months at -20 °C.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service