Skip to Content
Merck
All Photos(1)

Key Documents

SRP8029

Sigma-Aldrich

Flagellin from Salmonella typhimurium

>95% (SDS-PAGE)

Synonym(s):

FliC, Intelectin-1

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
$319.00

$319.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 2 weeks.

Request a Bulk Order

Select a Size

Change View
10 μG
$319.00

About This Item

CAS Number:
UNSPSC Code:
12352202
NACRES:
NA.32

$319.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 2 weeks.

Request a Bulk Order

biological source

Salmonella enterica subsp. enterica serovar Typhimurium str. LT2

Assay

>95% (SDS-PAGE)

form

lyophilized

mol wt

~50 kDa by SDS-PAGE

packaging

pkg of 10 μg

impurities

<0.01 EU/μg endotoxin, tested

color

white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

Salmonella enterica subsp. enterica serovar Typhimurium str. LT2 ... fliC (STM1959)

General description

FliC (flagellin) is the subunit protein which polymerizes to form the filaments of bacterial flagella. The protein has four-domains (D0, D1, D2, D3).[1]
The encoded flagellin filament contains 55 kDa monomers.[2]

Biochem/physiol Actions

FliC (flagellin) can activate the innate immune system by TLR5 (toll like receptor 5) and is an important proinflammatory molecule.[1][3] FliC also induces the production of antibodies and is identified as an antigen by CD4+ T cells.[4]
It also stimulates the production of proinflammatory mediators : nitric oxide, interleukin 1 (IL-1), IL-6, IL-8, and tumor necrosis factor α.[5]

Physical form

Lyophilized from PBS.

Reconstitution

Reconstitute with 100 μL deionized water.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mucosal administration of flagellin induces innate immunity in the mouse lung.
Honko A N and Mizel S B
Infection and Immunity, 72(11), 6676-6679 (2004)
In vivo, fliC expression by Salmonella enterica serovar Typhimurium is heterogeneous, regulated by ClpX, and anatomically restricted.
Cummings LA
Molecular Microbiology, 61, 795-809 (2006)
FliC-specific CD4+ T cell responses are restricted by bacterial regulation of antigen expression.
Cummings LA
Journal of Immunology, 174, 7929-7938 (2005)
Laís Rossi Perazza et al.
The Journal of nutrition, 150(10), 2673-2686 (2020-09-05)
Recent meta-analyses suggest that the consumption of fermented dairy products reduces type 2 diabetes and cardiovascular disease (CVD) risk, although the underlying mechanisms remain unclear. We evaluated whether dairy protein products modulated gut microbiota and cardiometabolic features in mouse models
Flagellin/TLR5 responses in epithelia reveal intertwined activation of inflammatory and apoptotic pathways.
Zeng H, et al.
American Journal of Physiology: Gastrointestinal and Liver Physiology, 290(1), G96-G108 (2006)

Questions

1–2 of 2 Questions  
  1. Hello! I wondered if this product is expressed from the host in its native folded state? I am interested in obtaining native folded flagellin... thanks for any insights you can provide in this matter!

    1 answer
    1. The structure of the material after purification is not assessed. While the native folding state cannot be guaranteed, the final purified flagellan can bind human and mouse TLR5 and activate TLR5 in cell culture.

      Helpful?

  2. Good morning, I would like to have the sequence of this protein and the host where it was produced. Thank yo and best regards

    1 answer
    1. This is native Flagellin purified from Salmonella typhimurium. The published sequence can be found here:
      https://www.uniprot.org/uniprotkb/P06179/entry

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service