Skip to Content
Merck
All Photos(1)

Key Documents

73996

Supelco

Lasalocid A sodium salt

analytical standard

Synonym(s):

Lasalocid sodium salt

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$112.00
5 MG
$210.00

$112.00


Estimated to ship onMarch 23, 2025


Request a Bulk Order

Select a Size

Change View
1 MG
$112.00
5 MG
$210.00

About This Item

Empirical Formula (Hill Notation):
C34H53NaO8
CAS Number:
Molecular Weight:
612.77
Beilstein:
3875309
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$112.00


Estimated to ship onMarch 23, 2025


Request a Bulk Order

grade

analytical standard

Assay

≥95.0% (HPLC)

form

powder

optical activity

[α]/D -28±3°, c = 1 in methanol

shelf life

limited shelf life, expiry date on the label

mp

175-185 °C
180 °C (dec.) (lit.)

application(s)

clinical testing

storage temp.

2-8°C

SMILES string

[Na+].CC[C@H]([C@H]1O[C@@](CC)(C[C@@H]1C)[C@H]2CC[C@](O)(CC)[C@H](C)O2)C(=O)[C@@H](C)[C@@H](O)[C@H](C)CCc3ccc(C)c(O)c3C([O-])=O

InChI

1S/C34H54O8.Na/c1-9-25(31-21(6)18-34(11-3,42-31)26-16-17-33(40,10-2)23(8)41-26)30(37)22(7)28(35)19(4)12-14-24-15-13-20(5)29(36)27(24)32(38)39;/h13,15,19,21-23,25-26,28,31,35-36,40H,9-12,14,16-18H2,1-8H3,(H,38,39);/q;+1/p-1

InChI key

RDHDUYAKDYQPEW-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Lasalocid is an antibacterial agent, belonging to the carboxylic polyether group of antibiotics, produced by the fermentation of Streptomyces lasaliensis.[1][2] It is commercially available as a sodium salt and is reported to be used as a coccidiostat in poultry.[3]

Application

Lasalocid A sodium salt may be used as an analytical standard for the quantification of lasalocid in the following:
  • Poultry feeds using liquid chromatography with ultraviolet detection (LC-UV).[4]
  • Animal liver and eggs using liquid chromatography coupled to tandem mass spectrometry (LC-MS-MS).[5]
  • Eggs using liquid chromatography-electrospray mass spectrometry (LC-EMS).[1]

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of lasalocid in eggs using liquid chromatography?electrospray mass spectrometry
Blanchflower JW and Kennedy GD
Analyst, 120(4), 1129-1132 (1995)
Chiral Drugs: Chemistry and Biological Action (2017)
Liquid chromatography with ultraviolet detection of lasalocid, monensin, salinomycin and narasin in poultry feeds using pre-column derivatization
Dusi G and Gamba V
Journal of Chromatography A, 835(1-2), 243-246 (1999)
The determination of 5 anticoccidial drugs (nicarbazin, lasalocid, monensin, salinomycin and narasin) in animal livers and eggs by liquid chromatography linked with tandem mass spectrometry (LC-MS-MS)
Matabudul KD, et al.
Analyst, 127(6), 760-768 (2002)
Code of Federal Regulations: Containing a Codification of Documents of General Applicability and Future Effect as of December 31, 1948, with Ancillaries and Index (2005)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service