Skip to Content
Merck
All Photos(4)

Key Documents

MSST0003

Sigma-Aldrich

SILuProt PTX3 Pentraxin-related protein PTX3 human

recombinant, expressed in HEK 293 cells, SIL MS Protein Standard, 13C- and 15N-labeled

Synonym(s):

SILu Prot Pentraxin-related protein

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
$992.00

$992.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

Select a Size

Change View
10 μG
$992.00

About This Item

UNSPSC Code:
23201100
NACRES:
NA.12

$992.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

biological source

human

Quality Level

recombinant

expressed in HEK 293 cells

tag

His tagged
V5 tagged

Assay

≥98% (SDS-PAGE)

form

lyophilized powder

technique(s)

mass spectrometry (MS): suitable

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... PTX3(5806)

General description

SILu Prot PTX3 is a recombinant, stable isotope-labeled human PTX3 which incorporates [13C6, 15N4]-Arginine and [13C6, 15N2]-Lysine. Expressed in human 293 cells, it is designed to be used as an internal standard for bioanalysis of PTX3 in mass-spectrometry. SILu Prot PTX3 is a recombinant glycosylated human protein expressed in human 293 cells. It is a homooctamer and a homodecamer consisting of 364 amino acids (monomer) with a calculated molecular weight of 40 kDa. It contains no tags.

Biochem/physiol Actions

Pentraxin 3 (PTX3) is a secreted glycosylated protein belonging to the pentraxin superfamily1. PTX3 is rapidly produced and released by several cell types, in particular by mononuclear phagocytes, dendritic cells (DCs), fibroblasts and endothelial cells in response to primary inflammatory signals (e.g., toll-like receptor [TLR]engagement, TNFα, IL-1β)1. PTX3 behaves as an acute phase response protein, as the blood levels of PTX3, low in normal conditions (about 25 ng/mL in the mouse, < 2 ng/mL in humans), increase rapidly (peaking at 6–8 h after induction)and dramatically (200–800 ng/mL)during endotoxic shock, sepsis and other inflammatory and infectious conditions, correlating with the severity of the disease. Under these conditions, PTX3 is a rapid marker for primary local activation of innate immunity and inflammation2-6. anti-apoptotic cell survival2, cell cycle regulation3, cell adhesion4, tissue remodeling5 and lipid transportation6. PTX3 gene expression in human endothelial cells is suppressed to a greater extent by pitavastatin than the expression of 6,000 other human genes that have been examined, suggesting that PTX3 may be a novel biomarker for inflammatory cardiovascular disease8.

Sequence

ENSDDYDLMYVNLDNEIDNGLHPTEDPTPCACGQEHSEWDKLFIMLENSQMRERMLLQATDDVLRGELQRLREELGRLAESLARPCAPGAPAEARLTSALDELLQATRDAGRRLARMEGAEAQRPEEAGRALAAVLEELRQTRADLHAVQGWAARSWLPAGCETAILFPMRSKKIFGSVHPVRPMRLESFSACIWVKATDVLNKTILFSYGTKRNPYEIQLYLSYQSIVFVVGGEENKLVAEAMVSLGRWTHLCGTWNSEEGLTSLWVNGELAATTVEMATGHIVPEGGILQIGQEKNGCCVGGGFDETLAFSGRLTGFNIWDSVLSNEEIRETGGAESCHIRGNIVGWGVTEIQPHGGAQYVS

Physical form

Supplied as a lyophilized powder containing phosphate buffered saline

Legal Information

This product is licensed under U.S. Patent No. 7,396,688 and foreign counterparts from E. I. du Pont de Nemours and Company. The purchase of this product conveys to the buyer the nontransferable right to use the purchased amount of the product for research and development only, including services for a third party for consideration. The buyer cannot sell or otherwise transfer this product, its components or materials made using this product or its components to a third party. Information about licenses for excluded uses is available from: E. I. du Pont de Nemours and Company; Attn: Associate Director, Commercial Development; DuPont Experimental Station E268; 200 Powdermill Rd.; Wilmington, DE 19803; 1-877-881-9787 (voice), 1-302-695-1437 (fax), [email protected].
SILu is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service