Skip to Content
Merck
All Photos(3)

Documents

R0514

Sigma-Aldrich

D-(+)-Raffinose pentahydrate

≥99% (HPLC), BioXtra

Synonym(s):

O-α-D-Galactopyranosyl-(1→6)-α-D-glucopyranosyl β-D-fructofuranoside, Melitose, Melitriose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H32O16 · 5H2O
CAS Number:
Molecular Weight:
594.51
Beilstein:
99541
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

product line

BioXtra

Assay

≥99% (HPLC)

form

powder

optical activity

[α]20/D +104 to +106°

technique(s)

HPLC: suitable

impurities

≤0.02% Phosphorus (P)
≤0.1% Insoluble matter

ign. residue

≤0.3%

color

white

solubility

H2O: 0.5 M, clear, colorless

anion traces

chloride (Cl-): ≤0.05%
sulfate (SO42-): ≤0.1%

cation traces

Al: ≤0.0005%
Ca: ≤0.002%
Cu: ≤0.0005%
Fe: ≤0.0005%
K: ≤0.2%
Mg: ≤0.002%
NH4+: ≤0.05%
Na: ≤0.005%
Pb: ≤0.001%
Zn: ≤0.0005%

SMILES string

O.O.O.O.O.OC[C@H]1O[C@H](OC[C@H]2O[C@H](O[C@]3(CO)O[C@H](CO)[C@@H](O)[C@@H]3O)[C@H](O)[C@@H](O)[C@@H]2O)[C@H](O)[C@@H](O)[C@H]1O

InChI

1S/C18H32O16.5H2O/c19-1-5-8(22)11(25)13(27)16(31-5)30-3-7-9(23)12(26)14(28)17(32-7)34-18(4-21)15(29)10(24)6(2-20)33-18;;;;;/h5-17,19-29H,1-4H2;5*1H2/t5-,6-,7-,8+,9-,10-,11+,12+,13-,14-,15+,16+,17-,18+;;;;;/m1...../s1

InChI key

BITMAWRCWSHCRW-PFQJHCPISA-N

Looking for similar products? Visit Product Comparison Guide

Application

D-(+)-Raffinose pentahydrate has been used as a fermentable sugar reference during in vitro colonic fermentation. It has also been used as a constituent of synthetic complete minus uracil media (Sc-ura).

Biochem/physiol Actions

Raffinose is a trisaccharide belonging to a family of oligosaccharides. It is extracted from various plants including beet sugar molasses, cottonseed meal and seed of food legumes. Raffinose improves cell proliferation and immune response. In addition, it also exhibits anti-oxidant and healing property. Raffinose is used as an additive in medicine, food and cosmetics.
Raffinose is a trisaccharide composed of galactose, fructose, and glucose. Raffinose is hydrolysed to D-galactose and sucrose by D-galactosidase.

Other Notes

To gain a comprehensive understanding of our extensive range of Disaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Probing Yeast Protein Microarrays for Protein-protein Interactions Using V5-epitope Tagged Fusion Protein Probes
Fasolo J and Snyder M
Genes & Development (2011)
Bioaccessibility during in vitro digestion and antiproliferative effect of bioactive compounds from Andean berry (Vaccinium meridionale Swartz) juice
Agudelo CD, et al.
Journal of Agricultural and Food Chemistry, 66(28), 7358-7366 (2018)
In vitro colonic fermentation of food ingredients isolated from Agave tequilana Weber var. azul applied on granola bars
Zamora Gasga V, et al.
LWT--Food Science and Technology, 60(2), 766-772 (2015)
Jidong Shen et al.
Bioengineered, 6(2), 99-105 (2015-02-20)
A glycoside hydrolase family 32 exo-inulinase gene was cloned from Arthrobacter sp. HJ7 isolated from saline soil located in Heijing town. The gene encodes an 892-residue polypeptide with a calculated mass of 95.1 kDa and a high total frequency of amino
The crystal structure of a trisaccharide, raffinose pentahydrate
Berman HM
Acta Crystallographica Section B, Structural Crystallography and Crystal Chemistry, 26(3), 290-299 (1970)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service