Skip to Content
Merck
All Photos(1)

Key Documents

MSST0007

Sigma-Aldrich

SILuProt CLU Clusterin human

recombinant, expressed in HEK 293 cells, SIL MS Protein Standard, 13C and 15N-labeled

Synonym(s):

Apolipoprotein J, Clusterin, Testosterone-repressed prostate message 2 (TRPM-2)

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
$992.00

$992.00


Estimated to ship onMay 26, 2025


Request a Bulk Order

Select a Size

Change View
10 μG
$992.00

About This Item

UNSPSC Code:
23201100
NACRES:
NA.12

$992.00


Estimated to ship onMay 26, 2025


Request a Bulk Order

biological source

human

Quality Level

recombinant

expressed in HEK 293 cells

tag

His tagged
V5 tagged

Assay

≥98% (SDS-PAGE)

form

lyophilized powder

potency

≥98% Heavy amino acids incorporation efficiency by MS

mol wt

56811.45

technique(s)

mass spectrometry (MS): suitable

suitability

suitable for mass spectrometry

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... CLU(1191)

Related Categories

General description

SILu Prot Clusterin is a recombinant, stable isotope-labeled human Clusterin which incorporates [13C6, 15N4]-Arginine and [13C6, 15N2]-Lysine. Expressed in human 293 cells, it is designed to be used as an internal standard for bioanalysis of Clusterin in mass-spectrometry. SIL Prot Clusterin is a heterodimer of 2 subunits (alpha and beta) consisting of 427 amino acids (including N-terminal polyhistidine and V5 tags), with a calculated molecular weight of 50 kDa.

Biochem/physiol Actions

Clusterin is a secreted glycosylated, 80-kDa disulfide-linked heterodimer of alpha and beta subunits (produced by internal cleavage). Clusterin is expressed in virtually all tissues and found in all human fluids. It is involved in numerous physiological processes important for carcinogenesis and tumor growth, including anti-apoptotic cell survival, cell cycle regulation, cell adhesion, tissue remodeling and lipid transportation. Clusterin also exists as a nuclear protein. The secreted form of Clusterin has extracellular chaperone and anti-apoptotic activities while the nuclear form acts as a proapoptotic factor.

Sequence

DQTVSDNELQEMSNQGSKYVNKEIQNAVNGVKQIKTLIEKTNEERKTLLSNLEEAKKKKEDALNETRESETKLKELPGVCNETMMALWEECKPCLKQTCMKFYARVCRSGSGLVGRQLEEFLNQSSPFYFWMNGDRIDSLLENDRQQTHMLDVMQDHFSRASSIIDELFQDRFFTREPQDTYHYLPFSLPHRRPHFFFPKSRIVRSLMPFSPYEPLNFHAMFQPFLEMIHEAQQAMDIHFHSPAFQHPPTEFIREGDDDRTVCREIRHNSTGCLRMKDQCDKCREILSVDCSTNNPSQAKLRRELDESLQVAERLTRKYNELLKSYQWKMLNTSSLLEQLNEQFNWVSRLANLTQGEDQYYLRVTTVASHTSDSDVPSGVTEVVVKLFDSDPITVTVPVEVSRKNPKFMETVAEKALQEYRKKHREESDPSRGPFEGKPIPNPLLGLDSTRTGHHHHHHHHGGQ

Physical form

Supplied as a lyophilized powder containing phosphate buffered saline.

Legal Information

SILu is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service