Skip to Content
Merck
All Photos(1)

Documents

MSST0064

Sigma-Aldrich

SILuProt Insulin, human

recombinant, expressed in Pichia pastoris, SIL MS Protein Standard, 15N -labeled

Synonym(s):

in situ Proximity Ligation Assay reagent, Insulin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23201100
NACRES:
NA.32

recombinant

expressed in Pichia pastoris

Quality Level

Assay

≥95% (HPLC)

form

dry pellets

potency

≥97% (Heavy amino acids incorporation efficiency by MS)

suitability

suitable for mass spectrometry (standard)

shipped in

ambient

storage temp.

−20°C

Gene Information

human ... INS(3630)

General description

SILu Prot Insulin is a recombinant, stable 15N isotope-labeled human Insulin Expressed in P. pastoris, it is designed to be used as an internal standard for bioanalysis of Insulin in mass-spectrometry. SILu Prot Insulin is a disulfate bonded hetero-dimer protein composed of two chains. Chain A of 21 amino acids and a thoretical amolecular mass of 2408.5; Chain B of 30 amino acids and a thoretical amolecular mass of 3468.7 (Note that Thr30 in chain B is not 15N labeled).

Biochem/physiol Actions

Insulin regulates the cellular uptake, utilization, and storage of glucose, amino acids, and fatty acids and inhibits the breakdown of glycogen, protein, and fat.

Sequence

A chain: GIVEQCCTSICSLYQLENYCN
B chain: FVNQHLCGSHLVEALYLVCGERGFFYTPKT

Physical form

Supplied as dried pellet from a solution containing 1% acetic acid.

Legal Information

This product is licensed under U.S. Patent No. 7,396,688 and foreign counterparts from E. I. du Pont de Nemours and Company. The purchase of this product conveys to the buyer the nontransferable right to use the purchased amount of the product for research and development only, including services for a third party for consideration. The buyer cannot sell or otherwise transfer this product, its components or materials made using this product or its components to a third party. Information about licenses for excluded uses is available from: E. I. du Pont de Nemours and Company; Attn: Associate Director, Commercial Development; DuPont Experimental Station E268; 200 Powdermill Rd.; Wilmington, DE 19803; 1-877-881-9787 (voice), 1-302-695-1437 (fax), licensing@dupont.com.
SILu is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service