Skip to Content
Merck
All Photos(1)

Key Documents

G019

Sigma-Aldrich

Muscimol hydrobromide

≥98% (HPLC), solid

Synonym(s):

5-Aminomethyl-3-hydroxyisoxazole hydrobromide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$179.00
10 MG
$305.00
25 MG
$605.00
100 MG
$2,020.00

$179.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$179.00
10 MG
$305.00
25 MG
$605.00
100 MG
$2,020.00

About This Item

Empirical Formula (Hill Notation):
C4H6N2O2 · HBr
CAS Number:
Molecular Weight:
195.01
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$179.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

solid

color

off-white to tan

solubility

H2O: >10 mg/mL

storage temp.

2-8°C

SMILES string

Br[H].NCc1cc(O)no1

InChI

1S/C4H6N2O2.BrH/c5-2-3-1-4(7)6-8-3;/h1H,2,5H2,(H,6,7);1H

InChI key

OPZOJWHOZRKYQX-UHFFFAOYSA-N

Biochem/physiol Actions

Muscimol hydrobromide is a GABAA receptor agonist.

Features and Benefits

This compound is a featured product for Neuroscience research. Click here to discover more featured Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the GABAA Receptors and GABAC Receptors pages of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Max F K Happel et al.
PloS one, 12(1), e0169461-e0169461 (2017-01-04)
Robust perception of auditory objects over a large range of sound intensities is a fundamental feature of the auditory system. However, firing characteristics of single neurons across the entire auditory system, like the frequency tuning, can change significantly with stimulus
Daniel H O'Connor et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 30(5), 1947-1967 (2010-02-05)
Linking activity in specific cell types with perception, cognition, and action, requires quantitative behavioral experiments in genetic model systems such as the mouse. In head-fixed primates, the combination of precise stimulus control, monitoring of motor output, and physiological recordings over
Elina A K Jacobs et al.
Current biology : CB, 30(24), 4944-4955 (2020-10-24)
In many behavioral tasks, cortex enters a desynchronized state where low-frequency fluctuations in population activity are suppressed. The precise behavioral correlates of desynchronization and its global organization are unclear. One hypothesis holds that desynchronization enhances stimulus coding in the relevant
Mohammed Z Ariffin et al.
Frontiers in neuroanatomy, 11, 79-79 (2017-10-03)
Cholinergic mechanisms in supramammillary nucleus (SuM), especially the lateral SuM (lSuM) modulates septo-hippocampal neural activity. The lSuM, as compared to the contiguous medial SuM (mSuM) has relatively dense projections to hippocampus and cingulate cortex (Cg). In the present study, we
Andrew Miri et al.
Neuron, 95(3), 683-696 (2017-07-25)
Blocking motor cortical output with lesions or pharmacological inactivation has identified movements that require motor cortex. Yet, when and how motor cortex influences muscle activity during movement execution remains unresolved. We addressed this ambiguity using measurement and perturbation of motor

Articles

DISCOVER Bioactive Small Molecules for Neuroscience

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service