Skip to Content
Merck
All Photos(1)

Key Documents

PHL89495

Muscimol

phyproof® Reference Substance

Synonym(s):

Agarine, Pantherine, 3-Hydroxy-5-aminomethyl-isoxazole, 5-Aminomethyl-3-hydroxy-isoxazole, 5-Aminomethyl-3-isoxazolol

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$759.00

$759.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Select a Size

Change View
10 MG
$759.00

About This Item

Empirical Formula (Hill Notation):
C4H6N2O2
CAS Number:
Molecular Weight:
114.10
Beilstein:
1618960
EC Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$759.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥98.0% (HPLC)

manufacturer/tradename

PhytoLab

application(s)

food and beverages

format

neat

storage temp.

−20°C

SMILES string

NCC1=CC(=O)NO1

InChI

1S/C4H6N2O2/c5-2-3-1-4(7)6-8-3/h1H,2,5H2,(H,6,7)

InChI key

ZJQHPWUVQPJPQT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Biochem/physiol Actions

GABAA receptor agonist.

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 2 Oral - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Laura A Bradfield et al.
Nature neuroscience, 23(10), 1194-1197 (2020-08-12)
The role of the hippocampus in goal-directed action is currently unclear; studies investigating this issue have produced contradictory results. Here we reconcile these contradictions by demonstrating that, in rats, goal-directed action relies on the dorsal hippocampus, but only transiently, immediately

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service