EP5895175007
Eppendorf® FA-6x250 rotor
for 6 x 250 mL tubes incl. QuickLock rotor cover, aerosol-tight, Centrifuge 5910 R/5920 R
Synonym(s):
Eppendorf Rotor
About This Item
Recommended Products
feature
for 6 x 250 mL tubes incl. QuickLock rotor cover, aerosol-tight, Centrifuge 5910 R/5920 R
packaging
case of 1 ea
manufacturer/tradename
Eppendorf® 5895175007
parameter
10,000 rpm speed
storage temp.
room temp
Related Categories
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service