Documents
HS120623
MagFuge High Speed Centrifuge Rotor
to hold, 6 x 5 mL tubes
About This Item
UNSPSC Code:
41103909
NACRES:
NB.78
feature
to hold 6 x 5 mL tubes
packaging
pkg of 1 ea
manufacturer/tradename
Heathrow Scientific 120623
parameter
12,500 rpm speed (max.)
9,783 × g max. RCF
compatibility
Heathrow Scientific HS10050
Legal Information
MagFuge is a trademark of Heathrow Scientific