Skip to Content
Merck

Skip To

B1908

Nalgene® centrifuge bottles, wide-mouth with sealing caps Style 3140

nominal capacity 250 mL

Synonym(s):

leak proof centrifuge bottle, Centrifuge containers, Laboratory centrifuge bottles, Nalgene 3140-0250, sample storage, spin bottle

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
4 ea
Check Cart for Availability
$328.00

About This Item

NACRES:
NB.11
UNSPSC Code:
41121703
Material:
polypropylene bottle, round bottle, screw top cap, transparent bottle, transparent polycarbonate (with sealing closure)
Parameter:
27500 × g max. RCF
Feature:
RCF 27,500 × g, with cap
Packaging:
pack of 4

$328.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

polypropylene bottle, round bottle, screw top cap, transparent bottle, transparent polycarbonate (with sealing closure)

joint

Joints threaded neck (46.5)

feature

RCF 27,500 × g, with cap

packaging

pack of 4

manufacturer/tradename

Nalgene® 3140-0250

parameter

27500 × g max. RCF

bottle diam. × L

61.5 mm × 135.1 mm

nominal capacity

250 mL

Features and Benefits

  • Nalgene® centrifuge bottles are used for large volume cell harvesting, pelleting, protein purification, and low-speed centrifugation of biological/chemical samples.
  • Translucent - for easy viewing of contents.
  • Strong with excellent mechanical properties.
  • Includes a polypropylene sealing cap closure with a silicone gasket that facilitates leak-proof assurance at maximum speeds and prevents sample loss.
  • Autoclavable.

Legal Information

Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
B2033Z353744Z353736
material

polypropylene bottle, screw top cap, transparent polycarbonate (with sealing closure), round bottle, transparent bottle

material

polypropylene bottle, round bottle, screw top cap, transparent bottle, transparent polycarbonate (with sealing closure)

material

flat bottom bottle, polypropylene copolymer (with sealing closure), round bottle, translucent bottle

material

flat bottom bottle, polypropylene copolymer (with sealing closure), translucent bottle

joint

Joints threaded neck (46.5)

joint

Joints threaded neck (52.6)

joint

Joints threaded neck (63)

joint

Joints threaded neck (58)

manufacturer/tradename

Nalgene® 3140-0250

manufacturer/tradename

Nalgene® 3140-0500

manufacturer/tradename

Nalgene® 3141-0500

manufacturer/tradename

Nalgene® 3141-0250

feature

RCF 27,500 × g, with cap

feature

RCF 13,700 × g, with cap

feature

RCF 13,700 × g, with cap

feature

RCF 27,500 × g, with cap

packaging

pack of 4

packaging

case of 24, pack of 4

packaging

pack of 4 ea

packaging

pack of 4 ea

parameter

27500 × g max. RCF

parameter

13700 × g max. RCF

parameter

13700 × g max. RCF

parameter

27500 × g max. RCF


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library