Skip to Content
Merck
All Photos(1)

Key Documents

CLS4612

Corning® microplate sealing tape

ELISA/General Incubation, polyester film, non-sterile

Synonym(s):

Microplate Sealing Tape

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122410
NACRES:
NB.13
Pricing and availability is not currently available.

material

polyester film

sterility

non-sterile

feature

piercable: no

packaging

pack of 500 ea

manufacturer/tradename

Corning 4612

parameter

-40-+120 °C temp. range

W × L

3.25 in. × 5.25 in.
8.26 cm × 13.34 cm

Looking for similar products? Visit Product Comparison Guide

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
CLS6524CLS3345CLS6569
material

polyester film

material

polyethylene film

material

white Rayon film (with acrylic)

material

aluminum film

manufacturer/tradename

Corning 4612

manufacturer/tradename

Corning 6524

manufacturer/tradename

Corning 3345

manufacturer/tradename

Corning 6569

sterility

non-sterile

sterility

non-sterile

sterility

sterile

sterility

non-sterile

packaging

pack of 500 ea

packaging

case of 100 ea

packaging

case of 500 ea

packaging

pack of 100

feature

piercable: no

feature

-

feature

-

feature

piercable

General description

  • Corning® microplate sealing tape is ideal for use in ELISA, general incubation, and other analytical assays.
  • The transparent tape ensures a tight seal to reduce evaporation/condensation and is used for both short- and long-term storage.
  • Uniform adhesive provides an effective well-to-well sealing across the entire plate to eliminate the edge effect in sensitive ELISA assays.
  • Suitable for sealing tissue culture microplates, for short-term storage incubation, transport, and the containment of solutions.

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service