Key Documents
Z743581
Titer Tops® sealing film for microplates
pack of 50
Synonym(s):
microplate sealing film, multiwell plate sealing film, plate seal
Select a Size
50 EA
$91.60
$91.60
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
50 EA
$91.60
About This Item
UNSPSC Code:
41103032
NACRES:
NB.17
$91.60
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
sterility
non-sterile
feature
prevents well-to-well contamination, sample spillage and evaporation
packaging
pack of 50
manufacturer/tradename
Diversified Biotech T-TOPS-50
General description
  • Durable and thick (4.4 mil) yet easily pierced with a pipette tip
  • Prevents well-to-well contamination, sample spillage and evaporation
  • Polyethylene film that provides excellent adhesion to plastic microplates
  • Non-cytotoxic, sterile, solvent resistant and waterproof
  • Stays solidly in place even when exposed to 100% humidity and extreme temperatures ranging from -40 °C to +100 °C (Not recommended for use in thermal cyclers)
  • Chemically resistant and thermally stable
  • Easily marked with any writing instrument
  • Two scored end taps for easy positioning
  • UV transparent down to 250 nm
Legal Information
Titer-Tops is a registered trademark of Diversified Biotech
Choose from one of the most recent versions:
Certificates of Analysis (COA)