CLS3585
Corning® 96 Well TC-Treated Microplates
size 96 wells, polystyrene, flat bottom, case of 50 (10 Bulk Packs of 5), sterile, lid (low evaporation)
Synonym(s):
96 well, cell culture plate, multiple well plates, multiwell plates, not treated, tissue culture plates
Select a Size
$108.00
Select a Size
About This Item
flat bottom
polystyrene
round clear wells
sterile
skirt
$108.00
Recommended Products
material
clear bottom
flat bottom
polystyrene
round clear wells
description
growth area .32 cm2 (low evaporation lid)
with low evaporation lid
sterility
sterile; γ-irradiated by SER-TAIN™ process
sterile
feature
lid (low evaporation)
skirt
packaging
case of 50 (10 Bulk Packs of 5)
manufacturer/tradename
Corning 3585
size
96 wells
well diam.
6.4 mm
well volume
360 μL
well working volume
75-200 μL
color
clear
binding type
Tissue Culture (TC)-treated surface
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service