CLS3512
Corning® Costar® TC-Treated Multiple Well Plates
size 12 wells, clear polystyrene plate, flat bottom, case of 100 (20 Bulk Packs of 5), sterile, lid
Synonym(s):
cell culture plate, multi well plates, multiple well plates, multiwell plates, tissue culture plates
Select a Size
$198.00
Estimated to ship onJune 02, 2025
Select a Size
About This Item
clear polystyrene plate
flat bottom
round clear wells
skirt
$198.00
Estimated to ship onJune 02, 2025
Recommended Products
material
clear bottom
clear polystyrene plate
flat bottom
round clear wells
description
growth area 3.8 cm2
sterility
sterile
feature
lid
skirt
packaging
case of 100 (20 Bulk Packs of 5)
manufacturer/tradename
Corning 3512
size
12 wells
well diam.
22.1 mm
well volume
6.9 mL
well working volume
0.76-1.14 mL
binding type
Tissue Culture (TC)-treated surface
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service